Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00820260_m1
          See other HAPLN3 GE Assays ›
          Gene Symbol:
          HAPLN3
          Gene Name:
          hyaluronan and proteoglycan link protein 3
          Gene Aliases:
          EXLD1, HsT19883
          Chromosome Location:
          Chr.15: 88877285 - 88895626 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00820260_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HAPLN3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001307952.1 NP_001294881.1 3 - 4 480 96
          NM_178232.3 NP_839946.1 2 - 3 341 96
          XM_011521261.1 XP_011519563.1 3 - 4 345 96
          XM_017021934.1 XP_016877423.1 3 - 4 456 96
          XM_017021935.1 XP_016877424.1 2 - 3 264 96
          XM_017021936.1 XP_016877425.1 3 - 4 451 96
          GenBank mRNA AK292055.1 - 2 - 3 264 96
          AK292103.1 - 2 - 3 342 96
          AY037161.1 - 2 - 3 229 96
          AY262759.1 - 2 - 3 254 96
          AY358500.1 - 2 - 3 226 96
          BC062320.1 - 3 - 4 451 5090960 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500010368.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the XM_017021935.1,XM_017021936.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HAPLN3

          Panther Classification:

          Molecular Function -

          extracellular matrix glycoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          cell adhesion
          central nervous system development
          extracellular matrix structural constituent
          hyaluronic acid binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000562281.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000562281.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000562281.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000562281.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000562281.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000562281.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000563808.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000563808.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000563808.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000563808.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 ENST00000563808.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500178061.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500178061.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500178061.hg.1 TC1500010368.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500178061.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500178061.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000562281.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000562281.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000562281.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000562281.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000562281.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000562281.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000563808.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000563808.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000563808.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000563808.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 ENST00000563808.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Clariom D Assay TC1500010368.hg.1 NM_178232.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500178058.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1500178058.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500178058.hg.1 ENST00000562281.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500178058.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500178058.hg.1 ENST00000563808.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500178058.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500178058.hg.1 NM_178232.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500178058.hg.1 TC1500010368.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500178058.hg.1 ENST00000359595.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1500178058.hg.1 ENST00000558770.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500178058.hg.1 ENST00000562281.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500178058.hg.1 ENST00000562889.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500178058.hg.1 ENST00000563808.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500178058.hg.1 NM_001307952.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500178058.hg.1 NM_178232.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 146996 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline