Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00826077_m1
          See other FGF18 GE Assays ›
          Gene Symbol:
          FGF18
          Gene Name:
          fibroblast growth factor 18
          Gene Aliases:
          FGF-18, ZFGF5
          Chromosome Location:
          Chr.5: 171419663 - 171457626 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00826077_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FGF18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003862.2 NP_003853.1 3 - 4 781 56
          GenBank mRNA AB007422.1 - 3 - 4 244 56
          AF075292.1 - 3 - 4 781 56
          AF211188.1 - 3 - 4 244 56
          AY358811.1 - 3 - 4 788 56
          BC006245.2 - 3 - 4 388 3948893 56
          BT019570.1 - 3 - 4 244 56
          BT019571.1 - 3 - 4 244 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500009424.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05000943.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FGF18

          Panther Classification:

          Molecular Function -

          growth factor intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          angiogenesis
          regulation of endothelial cell proliferation
          intramembranous ossification
          endochondral ossification
          chondrocyte development
          signal transduction
          cell-cell signaling
          positive regulation of cell proliferation
          fibroblast growth factor receptor signaling pathway
          anatomical structure morphogenesis
          regulation of phosphatidylinositol 3-kinase signaling
          peptidyl-tyrosine phosphorylation
          lung development
          positive regulation of vascular endothelial growth factor receptor signaling pathway
          positive regulation of chondrocyte differentiation
          phosphatidylinositol-3-phosphate biosynthetic process
          positive regulation of MAP kinase activity
          positive regulation of GTPase activity
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          positive regulation of ERK1 and ERK2 cascade
          regulation of sprouting angiogenesis
          positive regulation of endothelial cell chemotaxis to fibroblast growth factor
          protein tyrosine kinase activity
          Ras guanyl-nucleotide exchange factor activity
          type 1 fibroblast growth factor receptor binding
          type 2 fibroblast growth factor receptor binding
          growth factor activity
          1-phosphatidylinositol-3-kinase activity
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR05013791.hg.1 TC05000943.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05013791.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05013791.hg.1 NM_003862.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05013793.hg.1 TC05000943.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05013793.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05013793.hg.1 NM_003862.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05013792.hg.1 TC05000943.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05013792.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05013792.hg.1 NM_003862.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05013790.hg.1 TC05000943.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05013790.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05013790.hg.1 NM_003862.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05013794.hg.1 TC05000943.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05013794.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05013794.hg.1 NM_003862.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009424.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009424.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009424.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009424.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009424.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009424.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009424.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009424.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009424.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009424.hg.1 NM_003862.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009424.hg.1 NM_003862.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009424.hg.1 NM_003862.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009424.hg.1 NM_003862.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009424.hg.1 NM_003862.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009424.hg.1 NM_003862.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009424.hg.1 NM_003862.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009424.hg.1 NM_003862.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500009424.hg.1 NM_003862.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500166087.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500166087.hg.1 NM_003862.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500166087.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500166087.hg.1 NM_003862.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500166087.hg.1 TC0500009424.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500166088.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500166088.hg.1 NM_003862.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500166088.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500166088.hg.1 NM_003862.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500166088.hg.1 TC0500009424.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500166090.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500166090.hg.1 NM_003862.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500166090.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500166090.hg.1 NM_003862.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500166090.hg.1 TC0500009424.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500166091.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500166091.hg.1 NM_003862.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500166091.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500166091.hg.1 NM_003862.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500166091.hg.1 TC0500009424.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500166089.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500166089.hg.1 NM_003862.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500166089.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500166089.hg.1 NM_003862.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500166089.hg.1 TC0500009424.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500166092.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500166092.hg.1 NM_003862.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500166092.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500166092.hg.1 NM_003862.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500166092.hg.1 TC0500009424.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0500009424.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500009424.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500009424.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500009424.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500009424.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500009424.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500009424.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500009424.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500009424.hg.1 ENST00000274625.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500009424.hg.1 NM_003862.2 Refseq
          Affymetrix Human Clariom D Assay TC0500009424.hg.1 NM_003862.2 Refseq
          Affymetrix Human Clariom D Assay TC0500009424.hg.1 NM_003862.2 Refseq
          Affymetrix Human Clariom D Assay TC0500009424.hg.1 NM_003862.2 Refseq
          Affymetrix Human Clariom D Assay TC0500009424.hg.1 NM_003862.2 Refseq
          Affymetrix Human Clariom D Assay TC0500009424.hg.1 NM_003862.2 Refseq
          Affymetrix Human Clariom D Assay TC0500009424.hg.1 NM_003862.2 Refseq
          Affymetrix Human Clariom D Assay TC0500009424.hg.1 NM_003862.2 Refseq
          Affymetrix Human Clariom D Assay TC0500009424.hg.1 NM_003862.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline