| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000404843.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000404843.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000404843.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000404843.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000404843.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000404843.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000404843.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000404843.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000404843.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000404843.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000497092.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000497092.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000497092.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000497092.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
ENST00000497092.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02026598.hg.1 |
ENST00000272341.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02026598.hg.1 |
ENST00000336112.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02026598.hg.1 |
ENST00000404843.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02026598.hg.1 |
ENST00000435504.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02026598.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02026598.hg.1 |
TC02001663.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200225926.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200225926.hg.1 |
ENST00000404843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200225926.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200225926.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200225926.hg.1 |
ENST00000404843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200225926.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200225926.hg.1 |
TC0200016645.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000336112.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000404843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000404843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000404843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000404843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000404843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000404843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000404843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000404843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000404843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000404843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000497092.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000497092.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000497092.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000497092.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
ENST00000497092.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016645.hg.1 |
NM_018263.5 |
Refseq |