| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT HG-U133+ PM Array Plate |
216298_PM_at |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
217381_PM_s_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010787.hg.1 |
ENST00000390346.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010787.hg.1 |
ENST00000390346.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013540.hg.1 |
ENST00000390348.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013540.hg.1 |
ENST00000390348.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013540.hg.1 |
ENST00000390348.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010783.hg.1 |
ENST00000390344.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010783.hg.1 |
ENST00000390344.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010783.hg.1 |
ENST00000419915.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010783.hg.1 |
ENST00000419915.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010783.hg.1 |
ENST00000613175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010783.hg.1 |
ENST00000613175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010783.hg.1 |
ENST00000613175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010783.hg.1 |
ENST00000613175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010783.hg.1 |
ENST00000613175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010787.hg.1 |
ENST00000390346.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010787.hg.1 |
ENST00000390346.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013540.hg.1 |
ENST00000390348.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013540.hg.1 |
ENST00000390348.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013540.hg.1 |
ENST00000390348.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010783.hg.1 |
ENST00000390344.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010783.hg.1 |
ENST00000390344.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010783.hg.1 |
ENST00000419915.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010783.hg.1 |
ENST00000419915.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010783.hg.1 |
ENST00000613175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010783.hg.1 |
ENST00000613175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010783.hg.1 |
ENST00000613175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010783.hg.1 |
ENST00000613175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010783.hg.1 |
ENST00000613175.1 |
Ensembl |
| Human Genome U133 Plus 2.0 Array |
217374_PM_x_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
216298_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
217381_PM_s_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
217381_s_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
217374_x_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
216298_PM_at |
- |
- |
| Human Genome U133A 2.0 Array |
217374_PM_x_at |
- |
- |
| Human Genome U133A 2.0 Array |
217374_x_at |
- |
- |
| AffymetrixPathwork Diagnostics Pathchip |
pw537558 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
217381_s_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
216298_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
216298_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
217381_s_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
216298_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
216298_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
217381_s_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
216298_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
217381_s_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
217381_s_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
216298_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
217381_s_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
217381_s_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
217381_s_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
216298_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
217381_s_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
216298_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
216298_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
217381_s_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
217381_s_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
216298_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
216298_at1 |
- |
- |
| Human Genome U133 Set |
217374_x_at |
- |
- |
| Human Genome U133 Set |
217374_PM_x_at |
- |
- |