Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00853124_s1
          See other TAS2R46 GE Assays ›
          Gene Symbol:
          TAS2R46
          Gene Name:
          taste 2 receptor member 46
          Gene Aliases:
          T2R46, T2R54
          Chromosome Location:
          Chr.12: 11061365 - 11062294 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00853124_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TAS2R46
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_176887.2 NP_795368.2 1 543 79
          GenBank mRNA BC115401.2 - 1 543 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200009931.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12001240.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TAS2R46

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of bitter taste
          G-protein coupled receptor signaling pathway
          G-protein coupled receptor activity
          bitter taste receptor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622924 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16533299 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622900 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622911 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622918 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622913 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622905 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622891 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16533290 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622904 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622910 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622930 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622926 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622899 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622909 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622892 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P376114 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P376114 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 24931 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 28457 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P376114 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P376114 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533267 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622905 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622892 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622930 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622915 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622882 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622926 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622901 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622913 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533265 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533261 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533290 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622911 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622925 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622921 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622891 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533248 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622917 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622924 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622927 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622888 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622909 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622928 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622899 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533290 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622892 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622913 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622921 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622925 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533267 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622924 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622909 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622905 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622915 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622930 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533261 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622899 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622901 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622927 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622926 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533248 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622888 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622911 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622891 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533265 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622882 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622928 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622917 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P376114 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P376114 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_176887_at - -
          Affymetrix Human Gene 1.0 ST Array 7961294 - -
          Affymetrix Human Gene 1.1 ST Array 7961293 - -
          Affymetrix Human Genome U219 Array 11742393_PM_at - -
          Affymetrix Human Genome U219 Array 11742394_x_at - -
          Affymetrix Human Genome U219 Array 11742393_at - -
          Affymetrix Human Genome U219 Array 11742394_PM_x_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR12016283.hg.1 ENST00000533467.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12016283.hg.1 NM_176887.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12016283.hg.1 TC12001240.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000531678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000531678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000531678.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000541175.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000541175.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000541175.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000610639.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 ENST00000610639.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 NM_001097643.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 NM_001097643.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 NM_001097643.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 NM_176884.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 NM_176884.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012763.hg.1 NM_176884.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200175032.hg.1 ENST00000533467.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200175032.hg.1 NM_176887.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200175032.hg.1 ENST00000533467.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200175032.hg.1 NM_176887.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200175032.hg.1 TC1200009931.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000531678.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000531678.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000531678.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000539585.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000541175.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000541175.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000541175.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000610639.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 ENST00000610639.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 NM_001097643.1 Refseq
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 NM_001097643.1 Refseq
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 NM_001097643.1 Refseq
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 NM_176884.2 Refseq
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 NM_176884.2 Refseq
          Affymetrix Human Clariom D Assay TC1200012763.hg.1 NM_176884.2 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047331 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047331 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 327672 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1811572 BC115401.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1811572 NM_176887.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1811572 BC115401.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1811572 NM_176887.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1811572 BC115401.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1811572 NM_176887.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1811572 BC115401.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1811572 NM_176887.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1811572 BC115401.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1811572 NM_176887.2 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_28882040-S BC115401.2 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_28882040-S NM_176887.2 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_4048-S - -
          Illumina human-6 v1.0 expression beadchip 0002370592 - -
          Illumina humanRef-8 v1.0 expression beadchip 0002370592 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1811572 BC115401.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1811572 NM_176887.2 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1811572 BC115401.2 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1811572 NM_176887.2 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_4048 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_4048 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1811572 BC115401.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1811572 NM_176887.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1811572 BC115401.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1811572 NM_176887.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1811572 BC115401.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1811572 NM_176887.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1811572 BC115401.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1811572 NM_176887.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1811572 BC115401.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1811572 NM_176887.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline