Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00853130_s1
          See other TAS2R19 GE Assays ›
          Gene Symbol:
          TAS2R19
          Gene Name:
          taste 2 receptor member 19
          Gene Aliases:
          MSTP058, T2R19, T2R23, T2R48, TAS2R23, TAS2R48
          Chromosome Location:
          Chr.12: 11021619 - 11022620 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00853130_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TAS2R19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_176888.2 NP_795369.1 1 307 119
          GenBank mRNA BC101804.1 - 1 307 8069310 119
          BC111998.1 - 1 319 8327466 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200009928.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12001238.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TAS2R19

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of bitter taste
          G-protein coupled receptor signaling pathway
          G-protein coupled receptor activity
          bitter taste receptor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622718 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622729 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622728 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622712 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16533053 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622701 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622697 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622704 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622705 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622714 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622695 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622692 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28622696 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3402188 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P340308 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P340312 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P340308 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 10240 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 29515 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 2373 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 58854 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P340308 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P340308 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3402188 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622696 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622704 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622718 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622694 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622692 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622697 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622729 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622712 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622714 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622701 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622709 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533057 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622705 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622722 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622721 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622703 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533026 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533053 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622714 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622712 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622692 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622696 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622703 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622701 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533026 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622705 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622718 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533053 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622709 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622721 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622697 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16533057 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622704 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622694 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622722 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28622729 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P340308 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P340312 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08190673 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08190678 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08190677 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08190679 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08190676 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08190674 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P08190672 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P340308 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P340312 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08190677 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16533032 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_176888_at - -
          Affymetrix Human Gene 1.0 ST Array 7961289 - -
          Affymetrix Human Gene 1.0 ST Array 7961288 - -
          Affymetrix Human Gene 1.0 ST Array 7961290 - -
          Affymetrix Human Gene 1.1 ST Array 7961287 - -
          Affymetrix HT HG-U133+ PM Array Plate 1570384_PM_at - -
          Affymetrix Human Genome U219 Array 11738326_at - -
          Affymetrix Human Genome U219 Array 11738326_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR12016274.hg.1 ENST00000390673.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12016274.hg.1 NM_176888.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12016274.hg.1 TC12001238.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1200009928.hg.1 ENST00000390673.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009928.hg.1 ENST00000390673.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009928.hg.1 ENST00000390673.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009928.hg.1 NM_176888.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200009928.hg.1 NM_176888.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200009928.hg.1 NM_176888.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200175028.hg.1 ENST00000390673.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200175028.hg.1 NM_176888.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200175028.hg.1 ENST00000390673.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200175028.hg.1 NM_176888.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200175028.hg.1 TC1200009928.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1200009928.hg.1 ENST00000390673.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200009928.hg.1 ENST00000390673.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200009928.hg.1 ENST00000390673.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200009928.hg.1 NM_176888.2 Refseq
          Affymetrix Human Clariom D Assay TC1200009928.hg.1 NM_176888.2 Refseq
          Affymetrix Human Clariom D Assay TC1200009928.hg.1 NM_176888.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200175029.hg.1 ENST00000390673.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200175029.hg.1 NM_176888.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200175029.hg.1 ENST00000390673.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200175029.hg.1 NM_176888.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200175029.hg.1 TC1200009928.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200175027.hg.1 ENST00000390673.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200175027.hg.1 NM_176888.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200175027.hg.1 ENST00000390673.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200175027.hg.1 NM_176888.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200175027.hg.1 TC1200009928.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 1570384_PM_at - -
          Human Genome U133 Plus 2.0 Array 1570384_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570384_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570384_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570384_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570384_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570384_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570384_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570384_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570384_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570384_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570384_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570384_at8 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047329 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047329 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 707408 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1694485 BC111998.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1694485 NM_176888.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1694485 BC101804.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1694485 BC111998.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1694485 NM_176888.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1694485 BC101804.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1694485 BC111998.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1694485 NM_176888.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1694485 BC101804.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1694485 BC111998.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1694485 NM_176888.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1694485 BC101804.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1694485 BC111998.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1694485 NM_176888.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1694485 BC101804.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_28882034-S BC111998.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_28882034-S NM_176888.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_28882034-S BC101804.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_10632-S - -
          Illumina human-6 v1.0 expression beadchip 0002120168 - -
          Illumina humanRef-8 v1.0 expression beadchip 0002120168 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1694485 BC111998.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1694485 NM_176888.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1694485 BC101804.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1694485 BC111998.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1694485 NM_176888.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1694485 BC101804.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 4290201 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_10632 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1694485 BC111998.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1694485 NM_176888.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1694485 BC101804.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1694485 BC111998.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1694485 NM_176888.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1694485 BC101804.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1694485 BC111998.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1694485 NM_176888.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1694485 BC101804.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1694485 BC111998.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1694485 NM_176888.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1694485 BC101804.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1694485 BC111998.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1694485 NM_176888.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1694485 BC101804.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline