Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00858490_g1
          See other VHRT GE Assays ›
          Gene Symbol:
          VHRT
          Gene Name:
          ventricular heart development associated lncRNA
          Gene Aliases:
          LINC01405, MASCC1
          Chromosome Location:
          Chr.12: 110936602 - 110937448 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00858490_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: VHRT
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_036513.1 1 - 2 96 76
          GenBank mRNA AJ276555.1 - 1 - 2 96 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200008856.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12000862.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AJ276555.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613775 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29048307 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613769 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613781 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613813 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29048324 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613788 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29048319 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613795 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613774 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613783 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613797 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613772 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613794 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613796 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613778 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613800 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29048311 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613787 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613768 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613804 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613776 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613792 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29048305 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613808 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16591591 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613805 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29048272 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29048323 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613782 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613815 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29048313 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08613802 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P211248 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 8780 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 55041 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P211248 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P211248 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P02702052 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 93038 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02702052 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.1 ST Array 7958724 - -
          Affymetrix HT HG-U133+ PM Array Plate 230195_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000331096.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000331096.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000547607.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000547607.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000547607.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000547607.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 NR_036513.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008856.hg.1 NR_036513.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12011328.hg.1 ENST00000331096.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12011328.hg.1 NR_036513.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12011328.hg.1 TC12000862.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12011327.hg.1 ENST00000331096.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12011327.hg.1 ENST00000547607.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12011327.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12011327.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12011327.hg.1 NR_036513.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12011327.hg.1 TC12000862.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000331096.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000331096.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000547607.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000547607.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000547607.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000547607.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 NR_036513.1 Refseq
          Affymetrix Human Clariom D Assay TC1200008856.hg.1 NR_036513.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200165723.hg.1 ENST00000331096.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200165723.hg.1 ENST00000547607.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200165723.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200165723.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200165723.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200165723.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200165723.hg.1 NR_036513.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200165723.hg.1 ENST00000331096.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200165723.hg.1 ENST00000547607.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200165723.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200165723.hg.1 ENST00000548368.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200165723.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200165723.hg.1 ENST00000553177.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200165723.hg.1 NR_036513.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200165723.hg.1 TC1200008856.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200165724.hg.1 ENST00000331096.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200165724.hg.1 NR_036513.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200165724.hg.1 ENST00000331096.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200165724.hg.1 NR_036513.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200165724.hg.1 TC1200008856.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 230195_PM_at - -
          Human Genome U133 Plus 2.0 Array 230195_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230195_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230195_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230195_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230195_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230195_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230195_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230195_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230195_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230195_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230195_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230195_at3 - -
          Human Genome U95 Set 75047_PM_f_at - -
          Human Genome U95 Set 75047_f_at - -
          Human Genome U95 Set 78537_PM_f_at - -
          Human Genome U95 Set 78537_f_at - -
          Human Genome U133 Set 230195_at - -
          Human Genome U133 Set 230195_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 107730 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3291578 NR_036513.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3291578 AJ276555.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3291578 AW015651.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline