| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008261.hg.1 |
ENST00000419628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008261.hg.1 |
ENST00000419628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008261.hg.1 |
ENST00000419628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008261.hg.1 |
ENST00000419628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008261.hg.1 |
ENST00000419628.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008261.hg.1 |
NR_131192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008261.hg.1 |
NR_131192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008261.hg.1 |
NR_131192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008261.hg.1 |
NR_131192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008261.hg.1 |
NR_131192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100156009.hg.1 |
ENST00000419628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100156009.hg.1 |
NR_131192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100156009.hg.1 |
TC2100008261.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100156009.hg.1 |
ENST00000419628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100156009.hg.1 |
NR_131192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100156015.hg.1 |
NR_131192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100156015.hg.1 |
TC2100008261.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100156015.hg.1 |
NR_131192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100156011.hg.1 |
ENST00000419628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100156011.hg.1 |
NR_131192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100156011.hg.1 |
TC2100008261.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100156011.hg.1 |
ENST00000419628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100156011.hg.1 |
NR_131192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100156010.hg.1 |
ENST00000419628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100156010.hg.1 |
NR_131192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100156010.hg.1 |
TC2100008261.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100156010.hg.1 |
ENST00000419628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100156010.hg.1 |
NR_131192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100156013.hg.1 |
ENST00000419628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100156013.hg.1 |
NR_131192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100156013.hg.1 |
TC2100008261.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100156013.hg.1 |
ENST00000419628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100156013.hg.1 |
NR_131192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008261.hg.1 |
ENST00000419628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008261.hg.1 |
ENST00000419628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008261.hg.1 |
ENST00000419628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008261.hg.1 |
ENST00000419628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008261.hg.1 |
ENST00000419628.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008261.hg.1 |
NR_131192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008261.hg.1 |
NR_131192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008261.hg.1 |
NR_131192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008261.hg.1 |
NR_131192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008261.hg.1 |
NR_131192.1 |
Refseq |