Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00863277_g1
          See other YWHAQ GE Assays ›
          Gene Symbol:
          YWHAQ
          Gene Name:
          tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein theta
          Gene Aliases:
          14-3-3, 1C5, HS1
          Chromosome Location:
          Chr.2: 9583967 - 9631055 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00863277_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: YWHAQ
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_006826.3 NP_006817.1 2 - 3 492 154
          GenBank mRNA AF070556.1 - 1 - 2 81 154
          AK297624.1 - 3 - 4 325 154
          AK302208.1 - 2 - 3 198 154
          AK309462.1 - 3 - 4 321 154
          AK316158.1 - 3 - 4 320 154
          BC001197.1 - 429 154
          BC050601.1 - 1 - 2 305 6501365 154
          BC056867.1 - 2 - 3 427 6164592 154
          BC093019.1 - 2 - 3 434 30520472 154
          X56468.1 - 2 - 3 420 154
          X57347.1 - 2 - 3 395 154

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200011690.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02001560.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK302208.1,AK316158.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Human c-Myc and Apoptosis Human DNA Damage Induced 14-3-3 Sigma Signaling HUMAN INSULIN RECEPTOR PATHWAY HUMAN 14-3-3 INDUCED INTRACELLULAR SIGNALING HUMAN IGF1R SIGNALING 1700 Inventoried Human_c-Myc_and_Apoptosis Human_DNA_Damage_Induced_14-3-3_Sigma_Signaling Human_14-3-3_Induced_Intracellular_Signaling Human_IGF1R_Signaling Human_Insulin_Receptor_Pathway Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene YWHAQ

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting
          small GTPase mediated signal transduction
          substantia nigra development
          negative regulation of ion transmembrane transport
          negative regulation of transcription, DNA-templated
          membrane organization
          positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
          protein binding
          protein C-terminus binding
          protein domain specific binding
          ion channel binding
          protein N-terminus binding
          14-3-3 protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0300012639.hg.1 ENST00000508556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400009185.hg.1 ENST00000436268.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010350.hg.1 ENST00000490943.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000238081.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000238081.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000238081.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000238081.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000238081.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000238081.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000238081.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000238081.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000238081.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000238081.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000238081.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000238081.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000238081.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000381844.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000381844.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000381844.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000381844.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000381844.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000381844.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000381844.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000381844.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000381844.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000381844.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000381844.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000381844.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000381844.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000446619.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000446619.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000446619.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000446619.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000446619.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000446619.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000460093.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000460093.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000474715.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000474715.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000474715.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000474715.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000474715.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000474715.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 ENST00000474715.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 NM_006826.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 NM_006826.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 NM_006826.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 NM_006826.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 NM_006826.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 NM_006826.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 NM_006826.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 NM_006826.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 NM_006826.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 NM_006826.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 NM_006826.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 NM_006826.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 NM_006826.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011690.hg.1 NM_006826.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006511.hg.1 ENST00000454211.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006511.hg.1 ENST00000454211.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010350.hg.1 ENST00000490943.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300012639.hg.1 ENST00000508556.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400009185.hg.1 ENST00000436268.2 Ensembl
          Affymetrix Human Clariom D Assay TC0300006511.hg.1 ENST00000454211.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006511.hg.1 ENST00000454211.1 Ensembl
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 214185 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0