| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008439.hg.1 |
ENST00000561882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008439.hg.1 |
ENST00000561882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008439.hg.1 |
ENST00000561882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008439.hg.1 |
ENST00000561882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008439.hg.1 |
ENST00000561882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008439.hg.1 |
ENST00000561882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008439.hg.1 |
ENST00000561882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
ENST00000412626.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
ENST00000412626.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
ENST00000412626.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
ENST00000412626.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
ENST00000412626.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
ENST00000412626.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
ENST00000412626.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
ENST00000412626.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
ENST00000412626.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008439.hg.1 |
ENST00000561882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008439.hg.1 |
ENST00000561882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008439.hg.1 |
ENST00000561882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008439.hg.1 |
ENST00000561882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008439.hg.1 |
ENST00000561882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008439.hg.1 |
ENST00000561882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008439.hg.1 |
ENST00000561882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
ENST00000412626.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
ENST00000412626.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
ENST00000412626.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
ENST00000412626.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
ENST00000412626.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
ENST00000412626.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
ENST00000412626.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
ENST00000412626.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
ENST00000412626.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010219.hg.1 |
NR_002817.2 |
Refseq |