| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008972.hg.1 |
ENST00000420725.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008972.hg.1 |
ENST00000420725.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008972.hg.1 |
ENST00000420725.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008972.hg.1 |
NR_110390.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008972.hg.1 |
NR_110390.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008972.hg.1 |
NR_110390.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00161622.hg.1 |
ENST00000420725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00161622.hg.1 |
NR_110390.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00161622.hg.1 |
ENST00000420725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00161622.hg.1 |
NR_110390.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00161622.hg.1 |
TC0X00008972.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00008972.hg.1 |
ENST00000420725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008972.hg.1 |
ENST00000420725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008972.hg.1 |
ENST00000420725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008972.hg.1 |
NR_110390.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008972.hg.1 |
NR_110390.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008972.hg.1 |
NR_110390.1 |
Refseq |