| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000270879.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000270879.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000270879.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000270879.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000270879.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000270879.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000270879.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000270879.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000270879.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000354982.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000354982.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000354982.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000354982.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000354982.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000354982.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000354982.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000354982.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000481748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000481748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000481748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000481748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000481748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000481748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000498393.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000498393.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000498393.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000498393.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000498393.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000498393.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
ENST00000498393.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
NM_003665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
NM_003665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
NM_003665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
NM_003665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
NM_003665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
NM_003665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
NM_003665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
NM_003665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
NM_003665.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
NM_173452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
NM_173452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
NM_173452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
NM_173452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
NM_173452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
NM_173452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
NM_173452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013429.hg.1 |
NM_173452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199774.hg.1 |
ENST00000270879.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199774.hg.1 |
ENST00000354982.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199774.hg.1 |
ENST00000481748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199774.hg.1 |
NM_003665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199774.hg.1 |
NM_173452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199774.hg.1 |
ENST00000270879.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199774.hg.1 |
ENST00000354982.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199774.hg.1 |
ENST00000481748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199774.hg.1 |
NM_003665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199774.hg.1 |
NM_173452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199774.hg.1 |
TC0100013429.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000270879.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000270879.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000270879.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000270879.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000270879.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000270879.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000270879.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000270879.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000270879.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000354982.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000354982.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000354982.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000354982.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000354982.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000354982.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000354982.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000354982.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000481748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000481748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000481748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000481748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000481748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000481748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000498393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000498393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000498393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000498393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000498393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000498393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
ENST00000498393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
NM_003665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
NM_003665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
NM_003665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
NM_003665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
NM_003665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
NM_003665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
NM_003665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
NM_003665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
NM_003665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
NM_173452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
NM_173452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
NM_173452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
NM_173452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
NM_173452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
NM_173452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
NM_173452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013429.hg.1 |
NM_173452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199773.hg.1 |
ENST00000270879.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199773.hg.1 |
ENST00000354982.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199773.hg.1 |
ENST00000481748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199773.hg.1 |
NM_003665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199773.hg.1 |
NM_173452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199773.hg.1 |
ENST00000270879.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199773.hg.1 |
ENST00000354982.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199773.hg.1 |
ENST00000481748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199773.hg.1 |
NM_003665.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199773.hg.1 |
NM_173452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199773.hg.1 |
TC0100013429.hg.1 |
Clariom D TC |