| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT HG-U133+ PM Array Plate |
230953_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046437.hg.1 |
TC02002885.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046437.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046437.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046437.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046434.hg.1 |
TC02002885.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046434.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046434.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046434.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046436.hg.1 |
TC02002885.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046436.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046436.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046438.hg.1 |
TC02002885.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046438.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046438.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046438.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046438.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046435.hg.1 |
TC02002885.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046435.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046435.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046435.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046435.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
ENST00000389816.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
ENST00000389816.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
ENST00000389816.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
ENST00000389816.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
ENST00000421868.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
ENST00000421868.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
ENST00000421868.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
ENST00000421868.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
ENST00000421868.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
ENST00000449669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
ENST00000449669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
ENST00000449669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
ENST00000449669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016098.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218237.hg.1 |
ENST00000421868.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218237.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218237.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218237.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218237.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218237.hg.1 |
ENST00000421868.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218237.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218237.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218237.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218237.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218237.hg.1 |
TC0200016098.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200218236.hg.1 |
ENST00000389816.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218236.hg.1 |
ENST00000421868.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218236.hg.1 |
ENST00000449669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218236.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218236.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218236.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218236.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218236.hg.1 |
ENST00000389816.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218236.hg.1 |
ENST00000421868.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218236.hg.1 |
ENST00000449669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218236.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218236.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218236.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218236.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218236.hg.1 |
TC0200016098.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200218234.hg.1 |
ENST00000449669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218234.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218234.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218234.hg.1 |
ENST00000449669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218234.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218234.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218234.hg.1 |
TC0200016098.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
ENST00000389816.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
ENST00000389816.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
ENST00000389816.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
ENST00000389816.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
ENST00000421868.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
ENST00000421868.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
ENST00000421868.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
ENST00000421868.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
ENST00000421868.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
ENST00000449669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
ENST00000449669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
ENST00000449669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
ENST00000449669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016098.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218233.hg.1 |
ENST00000389816.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218233.hg.1 |
ENST00000421868.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218233.hg.1 |
ENST00000449669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218233.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218233.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218233.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218233.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218233.hg.1 |
ENST00000389816.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218233.hg.1 |
ENST00000421868.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218233.hg.1 |
ENST00000449669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218233.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218233.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218233.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218233.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218233.hg.1 |
TC0200016098.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200218231.hg.1 |
ENST00000421868.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218231.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218231.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218231.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218231.hg.1 |
ENST00000421868.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218231.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218231.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218231.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218231.hg.1 |
TC0200016098.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200218232.hg.1 |
ENST00000389816.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218232.hg.1 |
ENST00000421868.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218232.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218232.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218232.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218232.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218232.hg.1 |
ENST00000389816.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218232.hg.1 |
ENST00000421868.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218232.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218232.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218232.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218232.hg.1 |
NR_037696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218232.hg.1 |
TC0200016098.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200218235.hg.1 |
ENST00000389816.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218235.hg.1 |
ENST00000449669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218235.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218235.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218235.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218235.hg.1 |
ENST00000389816.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218235.hg.1 |
ENST00000449669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218235.hg.1 |
ENST00000450901.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200218235.hg.1 |
NR_037694.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218235.hg.1 |
NR_037695.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200218235.hg.1 |
TC0200016098.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
230953_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
230953_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
230953_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
230953_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
230953_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
230953_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
230953_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
230953_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
230953_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
230953_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
230953_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
230953_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
230953_at3 |
- |
- |
| Human Genome U95 Set |
76333_PM_at |
- |
- |
| Human Genome U95 Set |
76333_at |
- |
- |
| Human Genome U133 Set |
230953_at |
- |
- |
| Human Genome U133 Set |
230953_PM_at |
- |
- |