Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00894176_m1
          See other SIM2 GE Assays ›
          Gene Symbol:
          SIM2
          Gene Name:
          single-minded family bHLH transcription factor 2
          Gene Aliases:
          HMC13F06, HMC29C01, SIM, bHLHe15
          Chromosome Location:
          Chr.21: 36699117 - 36750219 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00894176_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SIM2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005069.4 NP_005060.1 7 - 8 1460 64
          NM_009586.3 NP_033664.2 7 - 8 1460 64
          XM_011529694.1 XP_011527996.1 6 - 7 1627 64
          XM_017028442.1 XP_016883931.1 7 - 8 1476 64
          GenBank mRNA AJ001858.1 - 6 - 7 658 64
          BC110444.2 - 7 - 8 872 40007501 64
          U80456.1 - 7 - 8 938 64
          U80457.1 - 7 - 8 938 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2100007056.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SIM2

          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          nervous system development
          embryonic pattern specification
          cell differentiation
          lung development
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000460783.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000460783.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000460783.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000460783.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000460783.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000483178.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000483178.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000483178.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 ENST00000483178.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Clariom D Assay PSR2100147687.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Clariom D Assay PSR2100147687.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100147687.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100147687.hg.1 NM_005069.5 Refseq
          Affymetrix Human Clariom D Assay PSR2100147687.hg.1 NM_009586.4 Refseq
          Affymetrix Human Clariom D Assay PSR2100147687.hg.1 TC2100007056.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100147687.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Clariom D Assay PSR2100147687.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100147687.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100147687.hg.1 NM_005069.5 Refseq
          Affymetrix Human Clariom D Assay PSR2100147687.hg.1 NM_009586.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000290399.10 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000431229.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000460783.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000460783.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000460783.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000460783.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000460783.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000481185.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000483178.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000483178.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000483178.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 ENST00000483178.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_005069.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_009586.4 Refseq
          Affymetrix Human Clariom D Assay TC2100007056.hg.1 NM_009586.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline