Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00896441_m1
          See other TET3 GE Assays ›
          Gene Symbol:
          TET3
          Gene Name:
          tet methylcytosine dioxygenase 3
          Gene Aliases:
          hCG_40738
          Chromosome Location:
          Chr.2: 73982036 - 74108176 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00896441_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TET3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001287491.1 NP_001274420.1 4 - 5 2584 57
          XM_005264187.3 XP_005264244.1 5 - 6 3104 57
          XM_011532682.2 XP_011530984.1 5 - 6 3104 57
          XM_011532683.2 XP_011530985.1 5 - 6 3413 57
          XM_011532684.2 XP_011530986.1 5 - 6 4845 57
          XM_011532685.2 XP_011530987.1 4 - 5 3047 57
          XM_011532686.2 XP_011530988.1 4 - 5 3047 57
          XM_011532687.2 XP_011530989.1 4 - 5 2386 57
          XM_011532688.2 XP_011530990.1 5 - 6 3104 57
          XM_011532689.2 XP_011530991.1 3 - 4 165 57
          XM_011532690.1 XP_011530992.1 2 - 3 222 57
          XM_017003566.1 XP_016859055.1 5 - 6 4944 57
          XM_017003567.1 XP_016859056.1 4 - 5 2416 57
          GenBank mRNA BC022243.1 - 1 - 2 40 4400878 57
          HQ220209.1 - 4 - 5 2584 57

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC022243.1, XM_011532689.2, XM_011532690.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein O-linked glycosylation
          DNA demethylation of male pronucleus
          positive regulation of transcription from RNA polymerase II promoter
          oxidation-reduction process
          DNA demethylation
          histone H3-K4 trimethylation
          DNA binding
          protein binding
          metal ion binding
          methylcytosine dioxygenase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000305799.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000305799.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000305799.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000305799.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000305799.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000305799.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000305799.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000305799.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000305799.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000305799.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000305799.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000305799.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000305799.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000305799.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000409262.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000409262.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000409262.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000409262.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000409262.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000409262.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000409262.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000409262.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000409262.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000409262.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000409262.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000409262.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000409262.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000409262.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000409262.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000409262.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000409262.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000475405.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000475405.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000475405.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000475405.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000496886.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 ENST00000496886.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 NM_001287491.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 NM_001287491.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 NM_001287491.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 NM_001287491.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 NM_001287491.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 NM_001287491.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 NM_001287491.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 NM_001287491.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 NM_001287491.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 NM_001287491.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 NM_001287491.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 NM_001287491.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 NM_001287491.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 NM_001287491.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 NM_001287491.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 NM_001287491.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008065.hg.1 NM_001287491.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 162432 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline