| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328502 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824174 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824125 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328514 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328509 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16826223 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328507 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328579 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824177 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824188 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824162 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824129 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824159 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824149 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824131 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328535 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328530 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824126 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824170 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328493 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824110 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16826230 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824169 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328583 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824123 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824140 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328506 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824127 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328549 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328588 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824137 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824132 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328570 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328585 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824164 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824141 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328581 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328560 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328489 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824156 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328494 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824161 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328525 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824118 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328569 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328516 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328488 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328526 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328476 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824130 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16826256 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328528 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824111 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328552 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824096 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328546 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328559 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824139 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824158 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328547 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328504 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824160 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328587 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328541 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16826248 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824154 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328540 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824102 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328524 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328565 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328527 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328564 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16826224 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328482 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16826250 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824106 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328499 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328563 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328542 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09824104 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P30328572 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
143444 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
48604 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
793665 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
349937 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
750298 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
390505 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
25590 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
209119 |
AL137763.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
209119 |
BC036890.2 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
209119 |
NM_021180.2 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
209119 |
NM_198173.1 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
209119 |
NM_198174.1 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
76241 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
121093 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3311740 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3230414 |
- |
- |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_0308 |
- |
- |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_0307 |
- |
- |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_0306 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
31354 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
8736 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P40498215 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P33788894 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_18_P20458447 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3230414 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3311740 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_33_P3230414 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_33_P3311740 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824125 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328539 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328538 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824140 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824123 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328524 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328514 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328527 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16826243 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328526 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824129 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824144 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824131 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824126 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824137 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328535 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824132 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328516 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328525 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824139 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328507 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328530 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328509 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824130 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824141 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328509 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328514 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328524 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824126 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328527 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824132 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824141 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824125 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16826243 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328539 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824123 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328525 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824131 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328526 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824144 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824139 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824140 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824129 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328530 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824137 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328538 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328507 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328516 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09824130 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P30328535 |
- |
- |
| Agilent Human Genome CGH Microarray 105A (G4412A) |
A_16_P03082684 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09824102 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09824129 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09824120 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09824121 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09824116 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09824108 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09824107 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09824130 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09824123 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09824137 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09824131 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09824133 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09824112 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09824109 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09824110 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09824113 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09824118 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09824134 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09824104 |
- |
- |
| Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09824106 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P09824129 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16826242 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16826240 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16826239 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16826241 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P09824131 |
- |
- |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P09824130 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P20344071 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P03082684 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
8833 |
- |
- |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
71315 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P03082684 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P12321714 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P40498215 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P12326658 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P33788993 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P03082691 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P20344071 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P03082693 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824131 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824125 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824134 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824146 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824158 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824179 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824110 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824116 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824167 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824187 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824153 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824181 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824175 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16826243 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824160 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824163 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824173 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824165 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824143 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824162 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824184 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824108 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824100 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824112 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16826245 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824182 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824176 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824106 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16826247 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16826248 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824186 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824170 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824140 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824115 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824137 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16826250 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16826259 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824138 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824129 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824142 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824133 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824169 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824149 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824104 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824178 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824130 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16826240 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824174 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824096 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824151 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824118 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824098 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824102 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16826253 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09824120 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P03082693 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P20344071 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P03082684 |
- |
- |