| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16014088.hg.1 |
ENST00000380897.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16014088.hg.1 |
ENST00000380898.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16014088.hg.1 |
ENST00000395724.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16014088.hg.1 |
ENST00000447459.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16014088.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16014088.hg.1 |
NM_144675.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16014088.hg.1 |
TC16000981.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000380897.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000380897.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000380897.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000380897.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000380897.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000380897.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000380897.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000380897.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000395724.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000395724.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000395724.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000395724.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000395724.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000395724.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000395724.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000395724.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000395724.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000562611.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000562611.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000562611.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000562611.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000562611.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000562611.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000562611.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000562611.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000569166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000569166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000569166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000569166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000569166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
ENST00000569166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
NM_144675.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
NM_144675.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
NM_144675.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
NM_144675.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
NM_144675.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
NM_144675.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
NM_144675.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
NM_144675.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009846.hg.1 |
NM_144675.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600175635.hg.1 |
ENST00000380897.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600175635.hg.1 |
ENST00000395724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600175635.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600175635.hg.1 |
ENST00000562611.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600175635.hg.1 |
ENST00000569166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600175635.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600175635.hg.1 |
NM_144675.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600175635.hg.1 |
ENST00000380897.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600175635.hg.1 |
ENST00000395724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600175635.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600175635.hg.1 |
ENST00000562611.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600175635.hg.1 |
ENST00000569166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600175635.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600175635.hg.1 |
NM_144675.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600175635.hg.1 |
TC1600009846.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000380897.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000380897.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000380897.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000380897.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000380897.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000380897.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000380897.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000380897.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000395724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000395724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000395724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000395724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000395724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000395724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000395724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000395724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000395724.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000447459.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000562611.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000562611.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000562611.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000562611.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000562611.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000562611.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000562611.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000562611.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000569166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000569166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000569166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000569166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000569166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
ENST00000569166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
NM_001109763.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
NM_144675.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
NM_144675.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
NM_144675.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
NM_144675.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
NM_144675.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
NM_144675.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
NM_144675.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
NM_144675.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009846.hg.1 |
NM_144675.2 |
Refseq |