Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00901870_m1
          See other NDUFAF6 GE Assays ›
          Gene Symbol:
          NDUFAF6
          Gene Name:
          NADH:ubiquinone oxidoreductase complex assembly factor 6
          Gene Aliases:
          C8orf38
          Chromosome Location:
          Chr.8: 94895271 - 95118496 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00901870_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NDUFAF6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_152416.3 NP_689629.2 6 - 7 742 65
          XM_005250789.2 XP_005250846.1 6 - 7 753 65
          XM_005250790.2 XP_005250847.1 7 - 8 853 65
          XM_005250791.2 XP_005250848.1 7 - 8 819 65
          XM_005250792.2 XP_005250849.1 7 - 8 823 65
          XM_005250793.2 XP_005250850.1 7 - 8 736 65
          XM_011516833.2 XP_011515135.1 11 - 12 1962 65
          XM_011516834.2 XP_011515136.1 10 - 11 1923 65
          XM_011516835.2 XP_011515137.1 10 - 11 1181 65
          XM_011516836.2 XP_011515138.1 10 - 11 1864 65
          XM_011516837.2 XP_011515139.1 11 - 12 1922 65
          XM_011516838.2 XP_011515140.1 10 - 11 1799 65
          XM_011516839.2 XP_011515141.1 9 - 10 1701 65
          XM_011516840.2 XP_011515142.1 10 - 11 1795 65
          XM_011516841.2 XP_011515143.1 10 - 11 1824 65
          XM_011516842.2 XP_011515144.1 9 - 10 1697 65
          XM_017013027.1 XP_016868516.1 10 - 11 1173 65
          XM_017013028.1 XP_016868517.1 9 - 10 1073 65
          XM_017013029.1 XP_016868518.1 8 - 9 1662 65
          XM_017013030.1 XP_016868519.1 8 - 9 1407 65
          XM_017013031.1 XP_016868520.1 9 - 10 1231 65
          XM_017013032.1 XP_016868521.1 7 - 8 853 65
          XM_017013033.1 XP_016868522.1 6 - 7 550 65
          XM_017013034.1 XP_016868523.1 5 - 6 523 65
          XM_017013035.1 XP_016868524.1 9 - 10 1275 65
          XM_017013036.1 XP_016868525.1 9 - 10 1607 65
          XM_017013037.1 XP_016868526.1 7 - 8 733 65
          XM_017013038.1 XP_016868527.1 8 - 9 802 65
          XM_017013039.1 XP_016868528.1 7 - 8 959 65
          GenBank mRNA AA625387.1 - 2 - 3 206 65
          AK298631.1 - 6 - 7 662 65
          AK301414.1 - 7 - 8 784 65
          AY444560.1 - 7 - 8 813 65
          BC028166.1 - 8 - 9 1075 65
          BF030925.1 - 2 - 3 253 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800012331.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08000579.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NDUFAF6

          Gene Ontology Categories:

          Function(s) Process(es)

          biosynthetic process
          mitochondrial respiratory chain complex I assembly
          transmembrane transport
          transferase activity
          transmembrane transporter activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000396124.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000396124.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000396124.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000396124.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000396124.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000396124.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000396124.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000396124.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000396124.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000396124.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000396124.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000396124.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000454358.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000454358.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000454358.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000454358.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000454358.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000454358.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000454358.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000454358.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000454358.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000454358.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000517976.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000517976.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000517976.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000517976.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000517976.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000517976.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000517976.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000517976.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000517976.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000517976.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000517976.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000517976.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000517976.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000517976.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000518258.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000518258.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000518258.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000518258.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000518258.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000518258.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000518258.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000518258.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000518258.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000518258.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000518258.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000518258.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000518258.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000518608.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000518608.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000518608.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000518608.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000518608.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000519804.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000519804.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000519804.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000519804.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000520632.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000520632.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000520632.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000520632.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000520632.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000520632.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000520632.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000520632.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000520632.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000522683.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000522683.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000522683.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000522683.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000522683.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000522683.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000523337.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000523337.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000523337.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000523337.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000523337.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000523337.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000523337.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 ENST00000523337.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 NM_152416.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 NM_152416.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 NM_152416.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 NM_152416.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 NM_152416.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 NM_152416.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 NM_152416.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 NM_152416.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 NM_152416.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 NM_152416.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 NM_152416.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012331.hg.1 NM_152416.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline