Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00902045_s1
          See other GABARAPL3 GE Assays ›
          Gene Symbol:
          GABARAPL3
          Gene Name:
          GABA type A receptor associated protein like 3 pseudogene
          Gene Aliases:
          ATG8D
          Chromosome Location:
          Chr.15: 90346527 - 90349447 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00902045_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GABARAPL3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028287.1 1 120 68
          GenBank mRNA AF180519.1 - 1 110 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500010435.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15001847.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AF180519.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          non-motor microtubule binding protein

          Gene Ontology Categories:

          Function(s)

          autophagosome assembly
          mitophagy
          cellular response to nitrogen starvation

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Oligo Microarray 4x44K 22365 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3812669 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09824118 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16826230 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30328504 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09824110 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09824111 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09824104 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30328494 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30328499 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30328502 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30328482 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09824099 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30328489 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09824113 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30328493 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09824102 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09824106 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30328506 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09824099 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09824104 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30328502 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30328504 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09824110 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09824113 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16826230 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30328489 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09824118 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30328506 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30328494 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30328499 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09824111 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09824102 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30328482 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30328493 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09824106 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.1 ST Array 7991395 - -
          Affymetrix HT HG-U133+ PM Array Plate 211457_PM_at - -
          Affymetrix Human Genome U219 Array 11760845_PM_x_at - -
          Affymetrix Human Genome U219 Array 11760845_x_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR15017923.hg.1 ENST00000412799.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15017923.hg.1 NR_028287.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15017923.hg.1 TC15001847.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1500008308.hg.1 ENST00000624044.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010435.hg.1 ENST00000412799.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010435.hg.1 NR_028287.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010435.hg.1 NR_028287.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010435.hg.1 NR_028287.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500178647.hg.1 NR_028287.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500178647.hg.1 NR_028287.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500178647.hg.1 TC1500010435.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1500008308.hg.1 ENST00000624044.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500178649.hg.1 NR_028287.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500178649.hg.1 NR_028287.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500178649.hg.1 TC1500010435.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500178648.hg.1 ENST00000412799.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1500178648.hg.1 NR_028287.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500178648.hg.1 ENST00000412799.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1500178648.hg.1 NR_028287.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500178648.hg.1 TC1500010435.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1500010435.hg.1 ENST00000412799.3 Ensembl
          Affymetrix Human Clariom D Assay TC1500010435.hg.1 NR_028287.1 Refseq
          Affymetrix Human Clariom D Assay TC1500010435.hg.1 NR_028287.1 Refseq
          Affymetrix Human Clariom D Assay TC1500010435.hg.1 NR_028287.1 Refseq
          Human Genome U133 Plus 2.0 Array 211458_PM_s_at - -
          Human Genome U133 Plus 2.0 Array 211458_s_at - -
          Human Genome U133A 2.0 Array 211458_s_at - -
          Human Genome U133A 2.0 Array 211458_PM_s_at - -
          Human Genome U133 Set 211458_s_at - -
          Human Genome U133 Set 211458_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052551 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052551 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 170375 - -
          ABI Human Genome Survey Microarray Version 2.0 185879 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline