Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00902143_m1
          See other TUBGCP3 GE Assays ›
          Gene Symbol:
          TUBGCP3
          Gene Name:
          tubulin gamma complex associated protein 3
          Gene Aliases:
          104p, GCP3, Grip104, SPBC98, Spc98, Spc98p
          Chromosome Location:
          Chr.13: 112485005 - 112606469 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00902143_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TUBGCP3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001286277.1 NP_001273206.1 5 - 6 726 70
          NM_001286278.1 NP_001273207.1 5 - 6 756 70
          NM_001286279.1 NP_001273208.1 5 - 6 756 70
          NM_006322.5 NP_006313.1 5 - 6 756 70
          XM_005268293.1 XP_005268350.1 5 - 6 751 70
          XM_011537462.2 XP_011535764.1 5 - 6 709 70
          XM_011537463.2 XP_011535765.1 6 - 7 685 70
          XM_011537464.2 XP_011535766.1 5 - 6 1489 70
          XM_017020323.1 XP_016875812.1 6 - 7 1103 70
          GenBank mRNA AF042378.1 - 5 - 6 635 70
          AJ003061.1 - 5 - 6 551 70
          AJ003062.1 - 5 - 6 551 70
          AK302537.1 - 5 - 6 673 70
          AK314785.1 - 5 - 6 756 70
          BC007763.2 - 5 - 6 706 70
          BC013781.1 - 5 - 6 696 3867019 70
          BC046634.2 - 5 - 6 739 5295831 70
          BT009772.1 - 5 - 6 551 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1300009856.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC13000887.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the XM_011537463.2, XM_011537464.2, XM_017020323.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TUBGCP3

          Panther Classification:

          Molecular Function -

          non-motor microtubule binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule nucleation
          meiotic nuclear division
          single fertilization
          cytoplasmic microtubule organization
          centrosome duplication
          interphase microtubule nucleation by interphase microtubule organizing center
          mitotic spindle assembly
          structural molecule activity
          structural constituent of cytoskeleton
          protein binding
          gamma-tubulin binding
          microtubule minus-end binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000462580.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000462580.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000462580.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000462580.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000462580.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000469302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000469302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000469302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000469302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000483532.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000483532.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000483532.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000483532.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000483532.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000483532.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000483532.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000611152.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000611152.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000611152.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000611152.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline