| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000563129.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000563129.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000563525.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000563525.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000563525.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000563525.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000563525.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000563525.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000563525.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000567975.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000567975.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000567975.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
ENST00000567975.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16013821.hg.1 |
ENST00000219689.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16013821.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16013821.hg.1 |
TC16000959.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000563129.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000563129.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000563525.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000563525.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000563525.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000563525.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000563525.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000563525.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000563525.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000567975.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000567975.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000567975.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
ENST00000567975.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009722.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600174861.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600174861.hg.1 |
NM_020718.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600174861.hg.1 |
TC1600009722.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600174861.hg.1 |
ENST00000219689.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600174861.hg.1 |
NM_020718.3 |
Refseq |