| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009856.hg.1 |
ENST00000302096.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009856.hg.1 |
ENST00000302096.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009856.hg.1 |
ENST00000302096.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009856.hg.1 |
ENST00000302096.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009856.hg.1 |
ENST00000308906.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009856.hg.1 |
ENST00000308906.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009856.hg.1 |
ENST00000308906.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009856.hg.1 |
ENST00000308906.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009856.hg.1 |
ENST00000308906.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009856.hg.1 |
NM_001007125.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009856.hg.1 |
NM_001007125.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009856.hg.1 |
NM_001007125.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009856.hg.1 |
NM_001007125.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009856.hg.1 |
NM_001007125.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014377.hg.1 |
ENST00000302096.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014377.hg.1 |
ENST00000308906.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014377.hg.1 |
NM_001007125.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014377.hg.1 |
TC20001047.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014378.hg.1 |
ENST00000302096.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014378.hg.1 |
ENST00000308906.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014378.hg.1 |
NM_001007125.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014378.hg.1 |
TC20001047.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC2000009856.hg.1 |
ENST00000302096.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009856.hg.1 |
ENST00000302096.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009856.hg.1 |
ENST00000302096.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009856.hg.1 |
ENST00000302096.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009856.hg.1 |
ENST00000308906.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009856.hg.1 |
ENST00000308906.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009856.hg.1 |
ENST00000308906.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009856.hg.1 |
ENST00000308906.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009856.hg.1 |
ENST00000308906.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009856.hg.1 |
NM_001007125.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009856.hg.1 |
NM_001007125.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009856.hg.1 |
NM_001007125.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009856.hg.1 |
NM_001007125.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009856.hg.1 |
NM_001007125.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000168862.hg.1 |
ENST00000302096.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168862.hg.1 |
ENST00000308906.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168862.hg.1 |
NM_001007125.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000168862.hg.1 |
ENST00000302096.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168862.hg.1 |
ENST00000308906.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168862.hg.1 |
NM_001007125.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000168862.hg.1 |
TC2000009856.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000168863.hg.1 |
ENST00000302096.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168863.hg.1 |
ENST00000308906.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168863.hg.1 |
NM_001007125.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000168863.hg.1 |
ENST00000302096.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168863.hg.1 |
ENST00000308906.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168863.hg.1 |
NM_001007125.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000168863.hg.1 |
TC2000009856.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000168864.hg.1 |
ENST00000302096.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168864.hg.1 |
ENST00000308906.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168864.hg.1 |
NM_001007125.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000168864.hg.1 |
ENST00000302096.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168864.hg.1 |
ENST00000308906.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168864.hg.1 |
NM_001007125.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000168864.hg.1 |
TC2000009856.hg.1 |
Clariom D TC |