| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000296581.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000296581.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000296581.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000296581.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000296581.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000296581.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000296581.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000502781.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000502781.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000502781.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000502781.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000502781.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000502781.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000502781.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000502781.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000502781.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000503982.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000503982.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000503982.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000504181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000504181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000504181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000504181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000504181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000504181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000507449.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000507449.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000507449.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000510331.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000510331.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000510331.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000515311.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000515311.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000515311.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000515311.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000515311.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
ENST00000515311.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
NM_007080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
NM_007080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
NM_007080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
NM_007080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
NM_007080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
NM_007080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008922.hg.1 |
NM_007080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010198.hg.1 |
ENST00000296581.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010198.hg.1 |
ENST00000502781.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010198.hg.1 |
ENST00000504181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010198.hg.1 |
ENST00000515311.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010198.hg.1 |
NM_007080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010198.hg.1 |
TC04000717.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000296581.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000296581.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000296581.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000296581.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000296581.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000296581.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000296581.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000502781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000502781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000502781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000502781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000502781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000502781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000502781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000502781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000502781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000503982.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000503982.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000503982.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000504181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000504181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000504181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000504181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000504181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000504181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000507449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000507449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000507449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000510331.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000510331.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000510331.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000515311.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000515311.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000515311.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000515311.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000515311.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
ENST00000515311.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
NM_007080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
NM_007080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
NM_007080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
NM_007080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
NM_007080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
NM_007080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008922.hg.1 |
NM_007080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400161317.hg.1 |
ENST00000296581.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161317.hg.1 |
ENST00000502781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161317.hg.1 |
ENST00000504181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161317.hg.1 |
ENST00000507449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161317.hg.1 |
ENST00000515311.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161317.hg.1 |
NM_007080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400161317.hg.1 |
ENST00000296581.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161317.hg.1 |
ENST00000502781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161317.hg.1 |
ENST00000504181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161317.hg.1 |
ENST00000507449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161317.hg.1 |
ENST00000515311.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161317.hg.1 |
NM_007080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400161317.hg.1 |
TC0400008922.hg.1 |
Clariom D TC |