| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10018878.hg.1 |
ENST00000308448.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10018878.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10018878.hg.1 |
ENST00000400215.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10018878.hg.1 |
ENST00000478142.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10018878.hg.1 |
ENST00000541004.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10018878.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10018878.hg.1 |
TC10001492.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007881.hg.1 |
ENST00000579735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007881.hg.1 |
ENST00000579735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007881.hg.1 |
ENST00000579735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007881.hg.1 |
ENST00000579735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000187509.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000187509.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000187509.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000187509.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000187509.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000187509.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000187509.hg.1 |
TC1000012575.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |