Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00907971_s1
          See other PFN3 GE Assays ›
          Gene Symbol:
          PFN3
          Gene Name:
          profilin 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 177400107 - 177400636 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00907971_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PFN3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001029886.2 NP_001025057.1 1 109 75
          GenBank mRNA BC132952.1 - 1 109 40146974 75
          BC132954.1 - 1 109 40146976 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500012970.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05002104.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PFN3

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          actin cytoskeleton organization
          regulation of actin filament polymerization
          positive regulation of actin filament bundle assembly
          actin binding
          lipid binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24669030 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24669040 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04487533 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24669028 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04487538 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24669038 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15768287 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P49997 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P49997 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 36359 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 51244 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P49997 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P49997 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04487533 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15768287 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24669040 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24669033 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04487536 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24669028 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24669039 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04487538 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24669038 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04487533 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04487538 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24669039 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24669038 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24669033 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24669040 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15768287 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24669028 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04487536 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P49997 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04487537 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04487533 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04487536 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15768292 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04487537 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15768290 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15768293 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04487536 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15768289 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15768295 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15768294 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15768291 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P49997 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 6618 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04487501 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04487533 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15768293 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04487564 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04487568 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04487560 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15768291 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04487494 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15768295 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04487504 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04487561 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04487572 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15768273 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04487493 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04487556 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04487537 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04487497 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04487574 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15768287 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04487559 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001029886_at - -
          Affymetrix Human Gene 1.0 ST Array 8116032 - -
          Affymetrix Human Gene 1.1 ST Array 8116031 - -
          Affymetrix Human Genome U219 Array 11733764_at - -
          Affymetrix Human Genome U219 Array 11733764_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR05029005.hg.1 TC05002104.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05029005.hg.1 ENST00000358571.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05029005.hg.1 NM_001029886.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500012970.hg.1 ENST00000358571.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500012970.hg.1 ENST00000358571.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500012970.hg.1 ENST00000358571.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500012970.hg.1 NM_001029886.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500012970.hg.1 NM_001029886.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500012970.hg.1 NM_001029886.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500190396.hg.1 ENST00000358571.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500190396.hg.1 NM_001029886.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500190396.hg.1 ENST00000358571.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500190396.hg.1 NM_001029886.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500190396.hg.1 TC0500012970.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0500012970.hg.1 ENST00000358571.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500012970.hg.1 ENST00000358571.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500012970.hg.1 ENST00000358571.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500012970.hg.1 NM_001029886.2 Refseq
          Affymetrix Human Clariom D Assay TC0500012970.hg.1 NM_001029886.2 Refseq
          Affymetrix Human Clariom D Assay TC0500012970.hg.1 NM_001029886.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500190398.hg.1 ENST00000358571.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500190398.hg.1 NM_001029886.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500190398.hg.1 ENST00000358571.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500190398.hg.1 NM_001029886.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500190398.hg.1 TC0500012970.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500190397.hg.1 ENST00000358571.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500190397.hg.1 NM_001029886.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500190397.hg.1 ENST00000358571.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500190397.hg.1 NM_001029886.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500190397.hg.1 TC0500012970.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A027741 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A027741 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 190449 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1718018 NM_001029886.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1718018 BC132952.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1718018 BC132954.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1718018 NM_001029886.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1718018 BC132952.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1718018 BC132954.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1718018 NM_001029886.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1718018 BC132952.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1718018 BC132954.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1718018 NM_001029886.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1718018 BC132952.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1718018 BC132954.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1718018 NM_001029886.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1718018 BC132952.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1718018 BC132954.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_5170-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1718018 NM_001029886.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1718018 BC132952.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1718018 BC132954.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1718018 NM_001029886.2 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1718018 BC132952.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1718018 BC132954.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 1990193 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_5170 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1718018 NM_001029886.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1718018 BC132952.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1718018 BC132954.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1718018 NM_001029886.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1718018 BC132952.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1718018 BC132954.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1718018 NM_001029886.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1718018 BC132952.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1718018 BC132954.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1718018 NM_001029886.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1718018 BC132952.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1718018 BC132954.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1718018 NM_001029886.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1718018 BC132952.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1718018 BC132954.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline