| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003062.hg.1 |
ENST00000375975.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003062.hg.1 |
ENST00000375980.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003062.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003062.hg.1 |
TC01000187.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
ENST00000445566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
ENST00000445566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
ENST00000445566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
ENST00000445566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
ENST00000445566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006982.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003061.hg.1 |
ENST00000375975.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003061.hg.1 |
ENST00000375980.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003061.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003061.hg.1 |
TC01000187.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003055.hg.1 |
ENST00000375980.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003055.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003055.hg.1 |
TC01000187.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003059.hg.1 |
ENST00000375975.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003059.hg.1 |
ENST00000375980.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003059.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003059.hg.1 |
TC01000187.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100148623.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148623.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100148623.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148623.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100148623.hg.1 |
TC0100006982.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100148628.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148628.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100148628.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148628.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100148628.hg.1 |
TC0100006982.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100148627.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148627.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100148627.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148627.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100148627.hg.1 |
TC0100006982.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100148630.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148630.hg.1 |
ENST00000445566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148630.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100148630.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148630.hg.1 |
ENST00000445566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148630.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100148630.hg.1 |
TC0100006982.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100148631.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148631.hg.1 |
ENST00000445566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148631.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100148631.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148631.hg.1 |
ENST00000445566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100148631.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100148631.hg.1 |
TC0100006982.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
ENST00000375980.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
ENST00000445566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
ENST00000445566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
ENST00000445566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
ENST00000445566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
ENST00000445566.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
NM_024329.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006982.hg.1 |
NM_024329.5 |
Refseq |