Search
Search

PCDHB8| Gene Symbol: PCDHB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019120.4 | NP_061993.3 | 1 | 2585 | 114 | |
| GenBank mRNA | AF152501.2 | - | 1 | 2340 | 114 | |
| BC136801.1 | - | 1 | 2585 | 9020791 | 114 | |
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04326375 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04326374 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24504775 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24504778 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24504776 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 242256 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 6690 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 348546 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 103844 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 340893 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 33644 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 418098 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 71513 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P41599 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P41599 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 18498 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 7056 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P31596124 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P41599 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P41599 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04326375 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24504776 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24504777 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24504784 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04326370 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24504793 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24504778 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24504781 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04326373 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24504793 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04326370 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24504781 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24504776 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24504784 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04326375 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24504778 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04326373 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24504777 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P41599 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04326370 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04326371 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04326368 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04326367 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04326375 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04326373 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04326365 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04326374 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P17324767 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P41599 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P15422955 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326429 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326377 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326407 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326351 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326400 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326331 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326416 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326335 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326365 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326421 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326405 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326328 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326402 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326433 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15747161 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326349 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326375 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326382 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326379 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326434 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326423 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326353 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326338 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326386 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326366 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326373 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326354 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326347 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326326 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326409 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326384 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326389 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326428 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326388 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15747141 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326419 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326342 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326356 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04326340 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_019120_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8108714 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8108715 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8108713 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 221319_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11741167_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11741167_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013243.hg.1 | ENST00000239444.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013243.hg.1 | ENST00000239444.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013243.hg.1 | ENST00000239444.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013243.hg.1 | ENST00000239444.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013243.hg.1 | NM_019120.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013243.hg.1 | NM_019120.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013243.hg.1 | NM_019120.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500194366.hg.1 | ENST00000239444.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500194366.hg.1 | NM_019120.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500194366.hg.1 | ENST00000239444.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500194366.hg.1 | NM_019120.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500194366.hg.1 | TC0500013243.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500013243.hg.1 | ENST00000239444.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013243.hg.1 | ENST00000239444.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013243.hg.1 | ENST00000239444.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013243.hg.1 | ENST00000239444.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013243.hg.1 | NM_019120.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013243.hg.1 | NM_019120.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013243.hg.1 | NM_019120.4 | Refseq |
| Human Genome U133 Plus 2.0 Array | 221319_at | - | - |
| Human Genome U133 Plus 2.0 Array | 221319_PM_at | - | - |
| Human Genome U133A 2.0 Array | 221319_PM_at | - | - |
| Human Genome U133A 2.0 Array | 221319_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_019120_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw532607 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221319_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221319_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221319_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221319_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221319_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221319_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221319_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221319_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221319_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221319_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221319_at9 | - | - |
| Human Genome U133 Set | 221319_PM_at | - | - |
| Human Genome U133 Set | 221319_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A028927 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A028927 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 169799 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1801051 | BC136801.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1801051 | NM_019120.3 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1801051 | AF152501.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1801051 | BC136801.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1801051 | NM_019120.3 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1801051 | AF152501.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1801051 | BC136801.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1801051 | NM_019120.3 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1801051 | AF152501.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1801051 | BC136801.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1801051 | NM_019120.3 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1801051 | AF152501.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1801051 | BC136801.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1801051 | NM_019120.3 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1801051 | AF152501.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_14195614-S | BC136801.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_14195614-S | NM_019120.3 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_14195614-S | AF152501.2 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_23560-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0002450020 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0002450020 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1801051 | BC136801.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1801051 | NM_019120.3 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1801051 | AF152501.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1801051 | BC136801.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1801051 | NM_019120.3 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1801051 | AF152501.2 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 1580411 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_23560 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1801051 | BC136801.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1801051 | NM_019120.3 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1801051 | AF152501.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1801051 | BC136801.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1801051 | NM_019120.3 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1801051 | AF152501.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1801051 | BC136801.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1801051 | NM_019120.3 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1801051 | AF152501.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1801051 | BC136801.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1801051 | NM_019120.3 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1801051 | AF152501.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1801051 | BC136801.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1801051 | NM_019120.3 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1801051 | AF152501.2 | human_transcriptome |