| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000506657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000506657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000506657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000506657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000506657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000506657.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000508252.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000508252.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000508252.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000511353.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000511353.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000511353.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000511353.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000512954.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000512954.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000512954.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000512954.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000514481.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000514481.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
ENST00000514481.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024806.hg.1 |
TC05001824.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024806.hg.1 |
ENST00000265191.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024806.hg.1 |
ENST00000508252.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024806.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000506657.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000506657.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000506657.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000506657.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000506657.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000506657.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000508252.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000508252.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000508252.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000511353.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000511353.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000511353.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000511353.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000512954.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000512954.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000512954.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000512954.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000514481.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000514481.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
ENST00000514481.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012160.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500184192.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184192.hg.1 |
ENST00000506657.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184192.hg.1 |
ENST00000508252.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184192.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500184192.hg.1 |
ENST00000265191.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184192.hg.1 |
ENST00000506657.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184192.hg.1 |
ENST00000508252.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500184192.hg.1 |
NM_003551.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500184192.hg.1 |
TC0500012160.hg.1 |
Clariom D TC |