| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17021852.hg.1 |
ENST00000006101.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17021852.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17021852.hg.1 |
TC17001633.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000579263.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000579263.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000579263.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000579263.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000579263.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000580174.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000580174.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000580174.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000580409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000580409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000580409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000580409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000580409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000580409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000581637.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000581637.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000581637.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000581637.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000581637.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000581637.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000581637.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000581756.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000581756.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000581756.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000581756.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000581756.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000581756.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000581756.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000581756.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000581756.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000583414.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000583414.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000583414.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000583414.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000583414.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000583414.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000584955.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000584955.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000584955.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000584955.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000584955.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000584955.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000584955.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000584955.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000584955.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000585182.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000585182.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000585182.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000585182.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000612370.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000612370.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000612370.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000612370.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000612370.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000612370.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700188255.hg.1 |
ENST00000580409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700188255.hg.1 |
ENST00000581637.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700188255.hg.1 |
ENST00000581756.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700188255.hg.1 |
ENST00000583414.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700188255.hg.1 |
ENST00000584955.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700188255.hg.1 |
ENST00000585182.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700188255.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700188255.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700188255.hg.1 |
ENST00000580409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700188255.hg.1 |
ENST00000581637.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700188255.hg.1 |
ENST00000581756.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700188255.hg.1 |
ENST00000583414.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700188255.hg.1 |
ENST00000584955.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700188255.hg.1 |
ENST00000585182.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700188255.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700188255.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700188255.hg.1 |
TC1700010973.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000579263.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000579263.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000579263.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000579263.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000579263.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000580174.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000580174.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000580174.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000580409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000580409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000580409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000580409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000580409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000580409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000581637.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000581637.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000581637.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000581637.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000581637.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000581637.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000581637.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000581756.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000581756.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000581756.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000581756.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000581756.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000581756.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000581756.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000581756.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000581756.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000583414.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000583414.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000583414.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000583414.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000583414.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000583414.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000584955.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000584955.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000584955.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000584955.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000584955.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000584955.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000584955.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000584955.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000584955.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000585182.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000585182.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000585182.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000585182.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000612370.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000612370.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000612370.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000612370.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000612370.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000612370.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
ENST00000621465.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010973.hg.1 |
NM_016429.3 |
Refseq |