Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00912660_m1
          See other ZC3HAV1 GE Assays ›
          Gene Symbol:
          ZC3HAV1
          Gene Name:
          zinc finger CCCH-type containing, antiviral 1
          Gene Aliases:
          ARTD13, FLB6421, PARP13, ZAP, ZC3H2, ZC3HDC2
          Chromosome Location:
          Chr.7: 139043519 - 139109720 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00912660_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ZC3HAV1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020119.3 NP_064504.2 2 - 3 832 69
          NM_024625.3 NP_078901.3 2 - 3 832 69
          XM_005250501.2 XP_005250558.1 2 - 3 833 69
          XM_011516412.2 XP_011514714.1 2 - 3 833 69
          GenBank mRNA AK055851.1 - 2 - 3 467 69
          AK292811.1 - 2 - 3 500 69
          BC025308.1 - 2 - 3 761 5418915 69
          BC033105.1 - 2 - 3 761 5418956 69
          BC040956.1 - 2 - 3 832 5531412 69
          BX571742.1 - 2 - 3 831 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700012761.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07001910.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ZC3HAV1

          Gene Ontology Categories:

          Function(s) Process(es)

          response to virus
          positive regulation of type I interferon production
          suppression by virus of host molecular function
          negative regulation of viral genome replication
          innate immune response
          regulation of defense response to virus by host
          defense response to virus
          positive regulation of mRNA catabolic process
          cellular response to exogenous dsRNA
          cell-cell adhesion
          NAD+ ADP-ribosyltransferase activity
          protein binding
          poly(A) RNA binding
          metal ion binding
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR07029641.hg.1 TC07001910.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07029641.hg.1 ENST00000242351.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07029641.hg.1 ENST00000464606.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07029641.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07029641.hg.1 ENST00000540247.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07029641.hg.1 NM_020119.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07029641.hg.1 NM_024625.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000460845.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000460845.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000460845.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000460845.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000460845.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000460845.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000460845.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000460845.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000460845.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000460845.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000460845.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000460845.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000460845.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000460845.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_020119.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Clariom D Assay TC0700012761.hg.1 NM_024625.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700192015.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0700192015.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700192015.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700192015.hg.1 NM_020119.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700192015.hg.1 NM_024625.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700192015.hg.1 ENST00000242351.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0700192015.hg.1 ENST00000464606.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700192015.hg.1 ENST00000471652.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700192015.hg.1 NM_020119.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700192015.hg.1 NM_024625.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700192015.hg.1 TC0700012761.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline