Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00915003_m1
          See other SLC24A3 GE Assays ›
          Gene Symbol:
          SLC24A3
          Gene Name:
          solute carrier family 24 member 3
          Gene Aliases:
          NCKX3
          Chromosome Location:
          Chr.20: 19212642 - 19722926 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00915003_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLC24A3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020689.4 NP_065740.2 2 - 3 475 71
          GenBank mRNA AF169257.2 - 2 - 3 217 71
          AF288087.1 - 2 - 3 471 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000006852.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000130.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SLC24A3

          Gene Ontology Categories:

          Function(s) Process(es)

          ion transport
          potassium ion transport
          sodium ion transport
          calcium ion transport
          cellular calcium ion homeostasis
          positive regulation of gene expression
          negative regulation of gene expression
          bone mineralization
          sodium ion transmembrane transport
          transmembrane transport
          calcium ion transmembrane transport
          potassium ion transmembrane transport
          transport across blood-brain barrier
          calcium channel activity
          calcium, potassium:sodium antiporter activity
          symporter activity
          antiporter activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20001664.hg.1 ENST00000328041.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20001664.hg.1 NM_020689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20001664.hg.1 TC20000130.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20001663.hg.1 ENST00000328041.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20001663.hg.1 NM_020689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20001663.hg.1 TC20000130.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2000008554.hg.1 ENST00000446849.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008554.hg.1 ENST00000446849.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20001665.hg.1 ENST00000328041.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20001665.hg.1 NM_020689.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20001665.hg.1 TC20000130.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR2000146878.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay PSR2000146878.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000146878.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000146878.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay PSR2000146878.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000146878.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000146878.hg.1 TC2000006852.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006852.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay TC2000008554.hg.1 ENST00000446849.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000008554.hg.1 ENST00000446849.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000146879.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay PSR2000146879.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000146879.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000146879.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay PSR2000146879.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000146879.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000146879.hg.1 TC2000006852.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000146882.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay PSR2000146882.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000146882.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000146882.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay PSR2000146882.hg.1 ENST00000613834.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000146882.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000146882.hg.1 TC2000006852.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000146877.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay PSR2000146877.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000146877.hg.1 ENST00000328041.10 Ensembl
          Affymetrix Human Clariom D Assay PSR2000146877.hg.1 NM_020689.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000146877.hg.1 TC2000006852.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline