| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000294521.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000294521.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000294521.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000294521.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000294521.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000294521.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000294521.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000373480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000373480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000373480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000373480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000373480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000373480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000373480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000373480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000373480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000373481.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000373481.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000373481.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000373481.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000373481.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000373481.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000373481.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000373481.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000401073.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000401073.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000401073.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000401073.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000401073.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000401073.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000401073.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000401073.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000401073.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000401073.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000468130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000468130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000468130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000468130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
ENST00000468130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_001198973.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_001198973.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_001198973.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_001198973.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_001198973.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_001198973.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_001198973.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_001198973.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_020888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_020888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_020888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_020888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_020888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_020888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_020888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_020888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007691.hg.1 |
NM_020888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01006724.hg.1 |
TC01000434.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01006724.hg.1 |
ENST00000373480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01006724.hg.1 |
ENST00000373481.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01006724.hg.1 |
ENST00000401073.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01006724.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01006724.hg.1 |
NM_020888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000294521.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000294521.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000294521.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000294521.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000294521.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000294521.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000294521.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000373480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000373480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000373480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000373480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000373480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000373480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000373480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000373480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000373480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000373481.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000373481.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000373481.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000373481.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000373481.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000373481.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000373481.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000373481.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000401073.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000401073.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000401073.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000401073.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000401073.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000401073.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000401073.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000401073.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000401073.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000401073.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000468130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000468130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000468130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000468130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
ENST00000468130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_001198973.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_001198973.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_001198973.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_001198973.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_001198973.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_001198973.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_001198973.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_001198973.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_020888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_020888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_020888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_020888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_020888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_020888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_020888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_020888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007691.hg.1 |
NM_020888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100154163.hg.1 |
ENST00000373480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100154163.hg.1 |
ENST00000373481.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100154163.hg.1 |
ENST00000401073.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100154163.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100154163.hg.1 |
NM_020888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100154163.hg.1 |
ENST00000373480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100154163.hg.1 |
ENST00000373481.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100154163.hg.1 |
ENST00000401073.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100154163.hg.1 |
NM_001198972.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100154163.hg.1 |
NM_020888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100154163.hg.1 |
TC0100007691.hg.1 |
Clariom D TC |