| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000593853.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000593853.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000593853.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000418929.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000593853.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000593853.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000593853.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
ENST00000615927.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008547.hg.1 |
NM_020719.2 |
Refseq |