Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00918595_m1
          See other PNOC GE Assays ›
          Gene Symbol:
          PNOC
          Gene Name:
          prepronociceptin
          Gene Aliases:
          N/OFQ, NOP, OFQ, PPNOC, ppN/OFQ
          Chromosome Location:
          Chr.8: 28316979 - 28343355 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00918595_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PNOC
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_006228.4 NP_006219.1 2 - 3 333 96
          XM_005273532.2 XP_005273589.1 2 - 3 420 96
          XM_011544559.2 XP_011542861.1 2 - 3 223 96
          GenBank mRNA AY335948.1 - 2 - 3 195 96
          BC034758.1 - 2 - 3 178 5195392 96
          BT019334.1 - 1 - 2 125 96
          CR541940.1 - 1 - 2 125 96
          CR541965.1 - 1 - 2 125 96
          U48263.1 - 2 - 3 336 96
          X97370.1 - 1 - 2 148 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800007132.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08000217.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PNOC

          Panther Classification:

          Molecular Function -

          neuropeptide

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          neuropeptide signaling pathway
          chemical synaptic transmission
          female pregnancy
          sensory perception
          opioid peptide activity
          neuropeptide hormone activity
          opioid receptor binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR08003167.hg.1 TC08000217.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08003167.hg.1 ENST00000301908.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08003167.hg.1 ENST00000522209.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08003167.hg.1 NM_006228.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800009978.hg.1 ENST00000521731.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009978.hg.1 ENST00000521731.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800149023.hg.1 ENST00000301908.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800149023.hg.1 ENST00000518479.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800149023.hg.1 ENST00000519592.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800149023.hg.1 ENST00000522209.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800149023.hg.1 NM_001284244.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800149023.hg.1 NM_006228.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800149023.hg.1 ENST00000301908.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800149023.hg.1 ENST00000518479.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800149023.hg.1 ENST00000519592.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800149023.hg.1 ENST00000522209.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800149023.hg.1 NM_001284244.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800149023.hg.1 NM_006228.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800149023.hg.1 TC0800007132.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0800009978.hg.1 ENST00000521731.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009978.hg.1 ENST00000521731.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800149022.hg.1 ENST00000301908.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800149022.hg.1 ENST00000518479.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800149022.hg.1 ENST00000519592.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800149022.hg.1 ENST00000522209.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800149022.hg.1 NM_001284244.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800149022.hg.1 NM_006228.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800149022.hg.1 ENST00000301908.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800149022.hg.1 ENST00000518479.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800149022.hg.1 ENST00000519592.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800149022.hg.1 ENST00000522209.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800149022.hg.1 NM_001284244.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800149022.hg.1 NM_006228.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800149022.hg.1 TC0800007132.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 182582 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline