| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000548256.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000548256.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000548256.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000548256.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000548256.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000551420.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000551420.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000551420.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000551420.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000551420.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000551420.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000552307.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000552307.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000552307.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
ENST00000552307.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007304.hg.1 |
ENST00000300209.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007304.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007304.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007304.hg.1 |
TC12003249.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007305.hg.1 |
ENST00000300209.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007305.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007305.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007305.hg.1 |
TC12003249.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007311.hg.1 |
ENST00000300209.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007311.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007311.hg.1 |
ENST00000548256.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007311.hg.1 |
ENST00000551420.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007311.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007311.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007311.hg.1 |
TC12003249.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007307.hg.1 |
ENST00000300209.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007307.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007307.hg.1 |
ENST00000552307.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007307.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007307.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007307.hg.1 |
TC12003249.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007312.hg.1 |
ENST00000300209.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007312.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007312.hg.1 |
ENST00000548256.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007312.hg.1 |
ENST00000551420.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007312.hg.1 |
ENST00000552307.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007312.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007312.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007312.hg.1 |
TC12003249.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1200200758.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200758.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200758.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200758.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200758.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200758.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200758.hg.1 |
TC1200012653.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200200763.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200763.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200763.hg.1 |
ENST00000548256.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200763.hg.1 |
ENST00000551420.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200763.hg.1 |
ENST00000552307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200763.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200763.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200763.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200763.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200763.hg.1 |
ENST00000548256.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200763.hg.1 |
ENST00000551420.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200763.hg.1 |
ENST00000552307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200763.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200763.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200763.hg.1 |
TC1200012653.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200200759.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200759.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200759.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200759.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200759.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200759.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200759.hg.1 |
TC1200012653.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200200761.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200761.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200761.hg.1 |
ENST00000552307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200761.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200761.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200761.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200761.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200761.hg.1 |
ENST00000552307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200761.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200761.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200761.hg.1 |
TC1200012653.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200200764.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200764.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200764.hg.1 |
ENST00000548256.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200764.hg.1 |
ENST00000551420.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200764.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200764.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200764.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200764.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200764.hg.1 |
ENST00000548256.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200764.hg.1 |
ENST00000551420.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200764.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200764.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200764.hg.1 |
TC1200012653.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000548256.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000548256.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000548256.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000548256.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000548256.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000551420.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000551420.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000551420.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000551420.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000551420.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000551420.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000552307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000552307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000552307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
ENST00000552307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012653.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200762.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200762.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200762.hg.1 |
ENST00000552307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200762.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200762.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200762.hg.1 |
ENST00000300209.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200762.hg.1 |
ENST00000333012.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200762.hg.1 |
ENST00000552307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200200762.hg.1 |
NM_015433.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200762.hg.1 |
NM_206914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200200762.hg.1 |
TC1200012653.hg.1 |
Clariom D TC |