| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008134.hg.1 |
ENST00000322055.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008134.hg.1 |
ENST00000370745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008134.hg.1 |
ENST00000370746.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008134.hg.1 |
ENST00000370750.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008134.hg.1 |
ENST00000484701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008134.hg.1 |
ENST00000545789.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008134.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008134.hg.1 |
TC06000681.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370748.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370748.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370748.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370748.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370748.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370748.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000370748.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000484701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000484701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000484701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000484701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000484701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
ENST00000484701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008135.hg.1 |
ENST00000322055.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008135.hg.1 |
ENST00000370746.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008135.hg.1 |
ENST00000370750.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008135.hg.1 |
ENST00000484701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008135.hg.1 |
ENST00000545789.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008135.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008135.hg.1 |
TC06000681.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370748.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370748.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370748.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370748.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370748.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370748.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000370748.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000484701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000484701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000484701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000484701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000484701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
ENST00000484701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008343.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600158199.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158199.hg.1 |
ENST00000484701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158199.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600158199.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158199.hg.1 |
ENST00000484701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158199.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600158199.hg.1 |
TC0600008343.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600158198.hg.1 |
ENST00000370745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158198.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158198.hg.1 |
ENST00000484701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158198.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600158198.hg.1 |
ENST00000370745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158198.hg.1 |
ENST00000370746.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158198.hg.1 |
ENST00000484701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158198.hg.1 |
NM_152731.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600158198.hg.1 |
TC0600008343.hg.1 |
Clariom D TC |