| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000470357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000470357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000470357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000470357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000470357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000487194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000487194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000487194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001593.hg.1 |
TC01000099.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001593.hg.1 |
ENST00000054666.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001593.hg.1 |
ENST00000470357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001593.hg.1 |
NM_004781.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011183.hg.1 |
ENST00000577089.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011183.hg.1 |
ENST00000577089.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011183.hg.1 |
ENST00000577089.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011183.hg.1 |
ENST00000577089.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000470357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000470357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000470357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000470357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000470357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000487194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000487194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000487194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011183.hg.1 |
ENST00000577089.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011183.hg.1 |
ENST00000577089.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011183.hg.1 |
ENST00000577089.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011183.hg.1 |
ENST00000577089.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146556.hg.1 |
ENST00000054666.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146556.hg.1 |
ENST00000470357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146556.hg.1 |
ENST00000487194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146556.hg.1 |
NM_004781.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146556.hg.1 |
ENST00000054666.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146556.hg.1 |
ENST00000470357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146556.hg.1 |
ENST00000487194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146556.hg.1 |
NM_004781.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146556.hg.1 |
TC0100006723.hg.1 |
Clariom D TC |