| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007203.hg.1 |
ENST00000292574.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007203.hg.1 |
ENST00000292574.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007203.hg.1 |
ENST00000292574.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007203.hg.1 |
ENST00000292574.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007203.hg.1 |
ENST00000292574.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007203.hg.1 |
ENST00000292574.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007203.hg.1 |
ENST00000292574.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007203.hg.1 |
ENST00000292574.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007203.hg.1 |
ENST00000292574.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007203.hg.1 |
NM_173482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007203.hg.1 |
NM_173482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007203.hg.1 |
NM_173482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007203.hg.1 |
NM_173482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007203.hg.1 |
NM_173482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007203.hg.1 |
NM_173482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007203.hg.1 |
NM_173482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007203.hg.1 |
NM_173482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007203.hg.1 |
NM_173482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003812.hg.1 |
ENST00000292574.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003812.hg.1 |
NM_173482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003812.hg.1 |
TC19000265.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900152032.hg.1 |
ENST00000292574.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152032.hg.1 |
NM_173482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900152032.hg.1 |
TC1900007203.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900152032.hg.1 |
ENST00000292574.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152032.hg.1 |
NM_173482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007203.hg.1 |
ENST00000292574.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007203.hg.1 |
ENST00000292574.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007203.hg.1 |
ENST00000292574.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007203.hg.1 |
ENST00000292574.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007203.hg.1 |
ENST00000292574.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007203.hg.1 |
ENST00000292574.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007203.hg.1 |
ENST00000292574.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007203.hg.1 |
ENST00000292574.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007203.hg.1 |
ENST00000292574.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007203.hg.1 |
NM_173482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007203.hg.1 |
NM_173482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007203.hg.1 |
NM_173482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007203.hg.1 |
NM_173482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007203.hg.1 |
NM_173482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007203.hg.1 |
NM_173482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007203.hg.1 |
NM_173482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007203.hg.1 |
NM_173482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007203.hg.1 |
NM_173482.2 |
Refseq |