Search
Search

CLCN3| Gene Symbol: CLCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243372.2 | NP_001230301.1 | 2 - 3 | 660 | 116 | |
| NM_001829.4 | NP_001820.2 | 2 - 3 | 660 | 116 | ||
| NM_173872.4 | NP_776297.2 | 2 - 3 | 660 | 116 | ||
| GenBank mRNA | AB019542.1 | - | 2 - 3 | 245 | 116 | |
| AF029346.1 | - | 1 - 2 | 158 | 116 | ||
| AF172729.1 | - | 1 - 2 | 158 | 116 | ||
| AK304362.1 | - | 2 - 3 | 651 | 116 | ||
| BC054877.1 | - | 2 - 3 | 575 | 116 | ||
| BX647119.1 | - | 2 - 3 | 667 | 116 | ||
| X78520.1 | - | 2 - 3 | 646 | 116 | ||
Set Membership: |
|