| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22008090.hg.1 |
ENST00000216075.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22008090.hg.1 |
ENST00000395732.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22008090.hg.1 |
ENST00000395733.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22008090.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22008090.hg.1 |
TC22000427.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200155462.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200155462.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200155462.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200155462.hg.1 |
ENST00000451761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200155462.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200155462.hg.1 |
TC2200007809.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200155462.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200155462.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200155462.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200155462.hg.1 |
ENST00000451761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200155462.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000216075.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000395733.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
ENST00000451761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007809.hg.1 |
NM_017584.5 |
Refseq |