| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000372499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000372499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000372499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000372499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000372499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000372499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000372499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000468134.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000468134.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000483375.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000483375.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000488655.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000488655.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000488655.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000488759.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000488759.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000488759.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000493194.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000493194.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000493690.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000493690.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000496424.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000496424.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000496424.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000496424.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000496424.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000496424.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000593699.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000593699.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000593699.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000593699.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000593699.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000593699.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000593699.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000593699.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000593817.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000593817.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000595936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000595936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000595936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000596228.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000596228.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000596228.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000596228.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000597823.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000597823.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000597823.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000597823.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000597823.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000598708.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000598708.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000598708.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000598708.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000611255.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000611255.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000611255.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000611255.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000611255.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000611255.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000611255.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000611255.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000611306.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000611306.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
ENST00000611306.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
NM_001305581.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
NM_001305581.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
NM_001305581.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
NM_001305581.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
NM_001305581.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
NM_001305581.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
NM_001305581.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
NM_032024.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
NM_032024.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
NM_032024.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
NM_032024.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
NM_032024.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
NM_032024.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
NM_032024.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
NR_131178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
NR_131178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
NR_131178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
NR_131178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
NR_131178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
NR_131178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
NR_131178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008099.hg.1 |
NR_131178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10005859.hg.1 |
ENST00000354343.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10005859.hg.1 |
ENST00000372499.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10005859.hg.1 |
NM_032024.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10005859.hg.1 |
TC10000488.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000372499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000372499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000372499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000372499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000372499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000372499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000372499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000468134.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000468134.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000483375.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000483375.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000488655.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000488655.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000488655.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000488759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000488759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000488759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000493194.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000493194.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000493690.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000493690.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000496424.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000496424.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000496424.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000496424.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000496424.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000496424.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000593699.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000593699.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000593699.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000593699.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000593699.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000593699.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000593699.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000593699.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000593817.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000593817.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000595936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000595936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000595936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000596228.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000596228.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000596228.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000596228.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000597823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000597823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000597823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000597823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000597823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000598708.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000598708.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000598708.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000598708.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000611255.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000611255.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000611255.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000611255.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000611255.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000611255.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000611255.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000611255.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000611306.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000611306.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
ENST00000611306.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
NM_001305581.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
NM_001305581.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
NM_001305581.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
NM_001305581.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
NM_001305581.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
NM_001305581.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
NM_001305581.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
NM_032024.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
NM_032024.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
NM_032024.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
NM_032024.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
NM_032024.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
NM_032024.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
NM_032024.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
NR_131178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
NR_131178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
NR_131178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
NR_131178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
NR_131178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
NR_131178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
NR_131178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008099.hg.1 |
NR_131178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000155047.hg.1 |
ENST00000372499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000155047.hg.1 |
ENST00000593699.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000155047.hg.1 |
ENST00000611255.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000155047.hg.1 |
ENST00000611306.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000155047.hg.1 |
NM_001305581.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000155047.hg.1 |
NM_032024.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000155047.hg.1 |
NR_131178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000155047.hg.1 |
ENST00000372499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000155047.hg.1 |
ENST00000593699.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000155047.hg.1 |
ENST00000611255.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000155047.hg.1 |
ENST00000611306.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000155047.hg.1 |
NM_001305581.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000155047.hg.1 |
NM_032024.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000155047.hg.1 |
NR_131178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000155047.hg.1 |
TC1000008099.hg.1 |
Clariom D TC |