Search
Search

OR4K15| Gene Symbol: OR4K15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005486.1 | NP_001005486.1 | 1 | 966 | 132 | |
| GenBank mRNA | BC140726.1 | - | 1 | 1041 | 9021594 | 132 |
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16682888 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29609351 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29609298 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29609304 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16682852 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29609313 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29609299 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29609330 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 139840 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3295710 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0671 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0670 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 21153 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P11918668 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3295710 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3295710 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16682852 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29609347 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09152861 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29609321 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29609330 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16682901 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29609329 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16682888 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29609311 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16682857 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09152860 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29609291 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16682913 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16682910 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16682902 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16682896 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16682877 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29609304 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29609298 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29609299 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29609313 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09152861 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29609304 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16682902 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29609291 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16682888 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09152860 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29609299 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29609347 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16682901 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16682877 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29609330 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29609313 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16682913 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29609329 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29609321 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29609311 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16682857 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16682896 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16682910 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16682852 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29609298 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09152857 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09152863 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09152856 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09152858 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09152861 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09152864 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09152859 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P09152862 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09152858 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09152864 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09152865 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09152859 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_001005486_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8020356 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7973019 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8020355 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8020354 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7973018 | - | - |
| Affymetrix Human Genome U219 Array | 11742312_at | - | - |
| Affymetrix Human Genome U219 Array | 11742312_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR14000165.hg.1 | ENST00000305051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14000165.hg.1 | NM_001005486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14000165.hg.1 | TC14000040.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006804.hg.1 | ENST00000578842.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006494.hg.1 | ENST00000305051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006494.hg.1 | ENST00000305051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006494.hg.1 | ENST00000305051.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006494.hg.1 | NM_001005486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007578.hg.1 | ENST00000431829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006803.hg.1 | ENST00000580232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006804.hg.1 | ENST00000578842.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006803.hg.1 | ENST00000580232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400144322.hg.1 | ENST00000305051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400144322.hg.1 | ENST00000305051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400144322.hg.1 | TC1400006494.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC2100007578.hg.1 | ENST00000431829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006494.hg.1 | ENST00000305051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006494.hg.1 | ENST00000305051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006494.hg.1 | ENST00000305051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006494.hg.1 | NM_001005486.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400144323.hg.1 | ENST00000305051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400144323.hg.1 | NM_001005486.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400144323.hg.1 | ENST00000305051.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400144323.hg.1 | NM_001005486.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400144323.hg.1 | TC1400006494.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051235 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051235 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 232987 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_2073115 | NM_001005486.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2073115 | BC140726.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1761294 | NM_001005486.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1761294 | NM_001005486.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1761294 | NM_001005486.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2073115 | NM_001005486.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2073115 | BC140726.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1761294 | NM_001005486.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2073115 | NM_001005486.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2073115 | BC140726.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1761294 | NM_001005486.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2073115 | NM_001005486.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2073115 | BC140726.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_13694-S | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1761294 | NM_001005486.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1761294 | NM_001005486.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 7040397 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_13694 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2073115 | NM_001005486.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2073115 | BC140726.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1761294 | NM_001005486.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2073115 | NM_001005486.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2073115 | BC140726.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2073115 | NM_001005486.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2073115 | BC140726.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1761294 | NM_001005486.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1761294 | NM_001005486.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2073115 | NM_001005486.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2073115 | BC140726.1 | human_transcriptome |