| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13004752.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13004752.hg.1 |
ENST00000397181.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13004752.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13004752.hg.1 |
TC13000421.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000471046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000471046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000471046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300154689.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300154689.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300154689.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300154689.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300154689.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300154689.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300154689.hg.1 |
TC1300008125.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000332556.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000471046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000471046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000471046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
ENST00000472564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008125.hg.1 |
NM_005561.3 |
Refseq |