Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00935807_m1
          See other LACC1 GE Assays ›
          Gene Symbol:
          LACC1
          Gene Name:
          laccase domain containing 1
          Gene Aliases:
          C13orf31, FAMIN, JUVAR
          Chromosome Location:
          Chr.13: 43879178 - 43893932 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00935807_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LACC1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001128303.2 NP_001121775.1 3 - 4 1334 84
          NM_001350638.2 NP_001337567.1 4 - 5 1143 84
          NM_001350639.2 NP_001337568.1 3 - 4 989 84
          NM_001350640.2 NP_001337569.1 3 - 4 948 84
          NM_001350641.2 NP_001337570.1 4 - 5 1152 84
          NM_001350642.2 NP_001337571.1 3 - 4 1126 84
          NM_001350643.2 NP_001337572.1 3 - 4 993 84
          NM_001350644.2 NP_001337573.1 3 - 4 689 84
          NM_001350645.2 NP_001337574.1 4 - 5 893 84
          NM_001350646.2 NP_001337575.1 3 - 4 739 84
          NM_001350647.2 NP_001337576.1 3 - 4 743 84
          NM_001350648.2 NP_001337577.1 3 - 4 743 84
          NM_153218.4 NP_694950.2 3 - 4 993 84
          XM_011534935.4 XP_011533237.1 4 - 5 1152 84
          XM_024449319.2 XP_024305087.1 3 - 4 1334 84
          XM_047430099.1 XP_047286055.1 4 - 5 1152 84
          XM_047430100.1 XP_047286056.1 3 - 4 855 84
          XM_047430101.1 XP_047286057.1 4 - 5 1152 84
          XM_047430102.1 XP_047286058.1 4 - 5 1538 84
          XM_047430103.1 XP_047286059.1 3 - 4 1126 84
          XM_047430104.1 XP_047286060.1 3 - 4 1334 84
          XM_047430105.1 XP_047286061.1 3 - 4 855 84
          XM_047430106.1 XP_047286062.1 4 - 5 1538 84
          XM_047430107.1 XP_047286063.1 3 - 4 1126 84
          XM_047430108.1 XP_047286064.1 3 - 4 948 84
          XM_047430109.1 XP_047286065.1 3 - 4 855 84
          GenBank mRNA AK096044.1 - 3 - 4 1064 84
          BC035749.1 - 3 - 4 1033 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1300009956.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC13000161.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_001350644.2, NM_001350645.2, NM_001350646.2, NM_001350647.2, NM_001350648.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LACC1

          Gene Ontology Categories:

          Function(s) Process(es)

          pattern recognition receptor signaling pathway
          immune system process
          positive regulation of cytokine production involved in immune response
          inflammatory response
          regulation of cellular pH
          regulation of defense response
          innate immune response
          regulation of inflammatory response
          nucleotide-binding oligomerization domain containing 2 signaling pathway
          regulation of purine nucleotide metabolic process
          adenosine deaminase activity
          purine-nucleoside phosphorylase activity
          copper ion binding
          protein binding
          oxidoreductase activity, acting on diphenols and related substances as donors, oxygen as acceptor
          transferase activity
          hydrolase activity
          S-methyl-5-thioadenosine phosphorylase activity
          metal ion binding
          guanosine phosphorylase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000325686.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000325686.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000325686.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000325686.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000325686.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000325686.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000325686.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000325686.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000325686.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000325686.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000325686.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000425906.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000425906.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000425906.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000441843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000441843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000441843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000441843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000441843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000441843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000441843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000441843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000441843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000441843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 ENST00000441843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 NM_001128303.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 NM_001128303.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 NM_001128303.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 NM_001128303.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 NM_001128303.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 NM_001128303.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 NM_001128303.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 NM_001128303.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 NM_001128303.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 NM_001128303.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 NM_001128303.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 NM_153218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 NM_153218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 NM_153218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 NM_153218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 NM_153218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 NM_153218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 NM_153218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 NM_153218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 NM_153218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 NM_153218.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009956.hg.1 NM_153218.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline