| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
ENST00000342745.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
ENST00000342745.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
ENST00000342745.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
ENST00000342745.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
ENST00000342745.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
ENST00000342745.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
ENST00000342745.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
ENST00000342745.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
ENST00000342745.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
ENST00000553497.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
ENST00000553497.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
ENST00000556993.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
ENST00000556993.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
ENST00000556993.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
ENST00000556993.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
NM_014734.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
NM_014734.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
NM_014734.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
NM_014734.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
NM_014734.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
NM_014734.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
NM_014734.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
NM_014734.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007558.hg.1 |
NM_014734.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14005039.hg.1 |
ENST00000342745.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14005039.hg.1 |
NM_014734.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14005039.hg.1 |
TC14000417.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
ENST00000342745.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
ENST00000342745.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
ENST00000342745.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
ENST00000342745.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
ENST00000342745.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
ENST00000342745.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
ENST00000342745.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
ENST00000342745.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
ENST00000342745.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
ENST00000553497.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
ENST00000553497.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
ENST00000556993.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
ENST00000556993.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
ENST00000556993.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
ENST00000556993.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
NM_014734.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
NM_014734.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
NM_014734.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
NM_014734.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
NM_014734.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
NM_014734.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
NM_014734.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
NM_014734.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007558.hg.1 |
NM_014734.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400152998.hg.1 |
ENST00000342745.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400152998.hg.1 |
NM_014734.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400152998.hg.1 |
ENST00000342745.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400152998.hg.1 |
NM_014734.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400152998.hg.1 |
TC1400007558.hg.1 |
Clariom D TC |