Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00938957_m1
          See other SNAP25 GE Assays ›
          Gene Symbol:
          SNAP25
          Gene Name:
          synaptosome associated protein 25
          Gene Aliases:
          CMS18, RIC-4, RIC4, SEC9, SNAP, SNAP-25, SUP, bA416N4.2, dJ1068F16.2
          Chromosome Location:
          Chr.20: 10218694 - 10307420 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00938957_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SNAP25
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001322902.1 NP_001309831.1 2 - 3 345 53
          NM_001322903.1 NP_001309832.1 3 - 4 441 53
          NM_001322904.1 NP_001309833.1 3 - 4 411 53
          NM_001322905.1 NP_001309834.1 3 - 4 374 53
          NM_001322906.1 NP_001309835.1 3 - 4 368 53
          NM_001322907.1 NP_001309836.1 4 - 5 569 53
          NM_001322908.1 NP_001309837.1 4 - 5 492 53
          NM_001322909.1 NP_001309838.1 3 - 4 359 53
          NM_001322910.1 NP_001309839.1 3 - 4 466 53
          NM_003081.4 NP_003072.2 2 - 3 314 53
          NM_130811.3 NP_570824.1 2 - 3 314 53
          XM_005260808.4 XP_005260865.1 2 - 3 392 53
          XM_017028021.1 XP_016883510.1 3 - 4 244 53
          XM_017028022.1 XP_016883511.1 3 - 4 336 53
          XM_017028023.1 XP_016883512.1 2 - 3 418 53
          GenBank mRNA AK094560.1 - 2 - 3 282 53
          AK098767.1 - 2 - 3 184 53
          AK223617.1 - 2 - 3 276 53
          AK289647.1 - 4 - 5 475 53
          AK314359.1 - 2 - 3 225 53
          BC010647.1 - 2 - 3 203 3867544 53
          BI552777.1 - 2 - 3 287 53
          BI602062.1 - 2 - 3 292 53
          BI604120.1 - 2 - 3 291 53
          BT019684.1 - 1 - 2 71 53
          BX647443.1 - 2 - 3 186 53
          D21267.1 - 2 - 3 276 53
          DA072367.1 - 2 - 3 314 53
          DA162693.1 - 2 - 3 418 53
          L19760.1 - 2 - 3 159 53
          L19761.1 - 2 - 3 159 53

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000006688.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000082.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SNAP25

          Panther Classification:

          Molecular Function -

          SNARE protein

          Gene Ontology Categories:

          Function(s) Process(es)

          neurotransmitter uptake
          chemical synaptic transmission
          neurotransmitter secretion
          locomotory behavior
          associative learning
          regulation of neuron projection development
          glutamate secretion
          synaptic vesicle exocytosis
          synaptic vesicle docking
          synaptic vesicle priming
          synaptic vesicle fusion to presynaptic active zone membrane
          regulation of insulin secretion
          long-term synaptic potentiation
          SNAP receptor activity
          protein binding
          syntaxin-1 binding
          calcium-dependent protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR20001178.hg.1 ENST00000254976.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20001178.hg.1 ENST00000304886.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20001178.hg.1 NM_003081.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20001178.hg.1 NM_130811.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20001178.hg.1 TC20000082.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000430336.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000430336.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000430336.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000430336.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000430336.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000430336.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000430336.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000492814.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000492814.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000492814.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000495883.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000495883.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000495883.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000495883.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000495883.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20001180.hg.1 ENST00000254976.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20001180.hg.1 ENST00000304886.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20001180.hg.1 NM_003081.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20001180.hg.1 NM_130811.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20001180.hg.1 TC20000082.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR2000145885.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2000145885.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2000145885.hg.1 ENST00000430336.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000145885.hg.1 ENST00000492814.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000145885.hg.1 NM_003081.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000145885.hg.1 NM_130811.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000145885.hg.1 TC2000006688.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000145885.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2000145885.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2000145885.hg.1 ENST00000430336.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000145885.hg.1 ENST00000492814.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000145885.hg.1 NM_003081.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000145885.hg.1 NM_130811.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000430336.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000430336.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000430336.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000430336.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000430336.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000430336.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000430336.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000492814.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000492814.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000492814.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000495883.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000495883.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000495883.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000495883.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000495883.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq
          Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline