Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00939736_m1
          See other EDA2R GE Assays ›
          Gene Symbol:
          EDA2R
          Gene Name:
          ectodysplasin A2 receptor
          Gene Aliases:
          EDA-A2R, EDAA2R, TNFRSF27, XEDAR
          Chromosome Location:
          Chr.X: 66595637 - 66639269 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00939736_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: EDA2R
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001199687.3 NP_001186616.2 2 - 3 160 103
          NM_001242310.1 NP_001229239.1 1 - 2 91 103
          NM_001324201.2 NP_001311130.2 2 - 3 160 103
          NM_001324206.2 NP_001311135.2 2 - 3 175 103
          NM_021783.5 NP_068555.2 2 - 3 175 103
          NR_136726.2 2 - 3 160 103
          NR_136727.2 2 - 3 175 103
          XM_011531001.2 XP_011529303.1 2 - 3 160 103
          XM_011531002.2 XP_011529304.1 2 - 3 175 103
          XM_011531006.3 XP_011529308.1 2 - 3 175 103
          XM_017029704.2 XP_016885193.1 2 - 3 175 103
          XM_017029706.2 XP_016885195.1 2 - 3 175 103
          XM_047442296.1 XP_047298252.1 2 - 3 175 103
          XM_047442299.1 XP_047298255.1 2 - 3 160 103
          XM_047442301.1 XP_047298257.1 2 - 3 160 103
          XM_047442303.1 XP_047298259.1 2 - 3 160 103
          XM_047442304.1 XP_047298260.1 2 - 3 175 103
          XM_047442305.1 XP_047298261.1 2 - 3 160 103
          XM_047442310.1 XP_047298266.1 2 - 3 160 103
          XM_047442311.1 XP_047298267.1 2 - 3 175 103
          XM_047442312.1 XP_047298268.1 2 - 3 160 103
          XM_047442313.1 XP_047298269.1 2 - 3 175 103
          XM_047442315.1 XP_047298271.1 2 - 3 160 103
          XM_047442317.1 XP_047298273.1 2 - 3 160 103
          XM_047442319.1 XP_047298275.1 2 - 3 160 103
          XM_047442321.1 XP_047298277.1 2 - 3 160 103
          XM_047442323.1 XP_047298279.1 2 - 3 160 103
          XM_047442324.1 XP_047298280.1 2 - 3 160 103
          XM_047442326.1 XP_047298282.1 2 - 3 175 103
          GenBank mRNA AF298812.1 - 1 - 2 81 103
          AK296264.1 - 2 - 3 141 103
          AK314881.1 - 2 - 3 157 103
          AY152724.1 - 1 - 2 81 103
          AY358735.1 - 1 - 2 84 103
          AY358736.1 - 1 - 2 84 103
          BC034919.1 - 2 - 3 138 103
          BP249106.1 - 2 - 3 157 103
          DA215848.1 - 2 - 3 190 104
          DA574050.1 - 2 - 3 141 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00009917.hg
          This assay targets the same location as HTA transcript cluster(s) TC0X001101.hg

          Additional Information:

          This assay is in the 5'UTR region of the NM_001324201.2, XM_047442317.1, XM_047442323.1, XM_047442326.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene EDA2R

          Gene Ontology Categories:

          Function(s) Process(es)

          epidermis development
          ectodermal cell differentiation
          cytokine-mediated signaling pathway
          cell differentiation
          tumor necrosis factor-mediated signaling pathway
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of JNK cascade
          intrinsic apoptotic signaling pathway by p53 class mediator
          transmembrane signaling receptor activity
          tumor necrosis factor-activated receptor activity
          protein binding
          signaling receptor activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR0X015030.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X015030.hg.1 TC0X001101.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR0X015023.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X015023.hg.1 ENST00000374719.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X015023.hg.1 ENST00000396050.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X015023.hg.1 ENST00000450752.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X015023.hg.1 ENST00000456230.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X015023.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X015023.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X015023.hg.1 NM_021783.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X015023.hg.1 TC0X001101.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR0X015029.hg.1 ENST00000456230.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X015029.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X015029.hg.1 TC0X001101.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00009917.hg.1 NM_021783.4 Refseq
          Affymetrix Human Clariom D Assay PSR0X00167757.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00167757.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00167757.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00167757.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00167757.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00167757.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00167757.hg.1 NM_021783.4 Refseq
          Affymetrix Human Clariom D Assay PSR0X00167757.hg.1 ENST00000253392.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00167757.hg.1 ENST00000374719.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00167757.hg.1 ENST00000396050.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00167757.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00167757.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00167757.hg.1 NM_001242310.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00167757.hg.1 NM_021783.4 Refseq
          Affymetrix Human Clariom D Assay PSR0X00167757.hg.1 TC0X00009917.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00167762.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00167762.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00167762.hg.1 ENST00000451436.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00167762.hg.1 NM_001199687.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00167762.hg.1 TC0X00009917.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 161767 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline