| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010227.hg.1 |
ENST00000368171.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010227.hg.1 |
ENST00000368171.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010227.hg.1 |
ENST00000368171.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010227.hg.1 |
ENST00000368171.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010227.hg.1 |
ENST00000368171.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010227.hg.1 |
ENST00000368171.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010227.hg.1 |
ENST00000368171.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010227.hg.1 |
ENST00000368171.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010227.hg.1 |
NM_001766.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010227.hg.1 |
NM_001766.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010227.hg.1 |
NM_001766.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010227.hg.1 |
NM_001766.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010227.hg.1 |
NM_001766.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010227.hg.1 |
NM_001766.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010227.hg.1 |
NM_001766.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010227.hg.1 |
NM_001766.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010227.hg.1 |
NM_001766.3 |
Refseq |