Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00941233_g1
          See other ORC6 GE Assays ›
          Gene Symbol:
          ORC6
          Gene Name:
          origin recognition complex subunit 6
          Gene Aliases:
          ORC6L
          Chromosome Location:
          Chr.16: 46689659 - 46698394 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00941233_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ORC6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_014321.4 NP_055136.1 3 - 4 409 108
          NR_037620.2 3 - 4 468 108
          XM_011522978.4 XP_011521280.1 3 - 4 409 108
          GenBank mRNA AF139658.1 - 3 - 4 410 108
          AK021946.1 - 3 - 4 383 108
          AK024019.1 - 3 - 4 416 108
          AK024077.1 - 3 - 4 409 108
          AK310633.1 - 3 - 4 445 108
          BC039032.1 - 3 - 4 402 108
          BC063565.1 - 3 - 4 391 108
          BG822640.1 - 3 - 4 393 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600007718.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC16000425.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ORC6

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA replication
          DNA-dependent DNA replication
          DNA replication initiation
          DNA binding
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1800006791.hg.1 ENST00000623071.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006791.hg.1 ENST00000623071.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006791.hg.1 ENST00000623071.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006791.hg.1 ENST00000624346.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009870.hg.1 ENST00000529323.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009870.hg.1 ENST00000529323.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009108.hg.1 ENST00000625200.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010748.hg.1 ENST00000624108.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000219097.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000219097.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000219097.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000219097.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000219097.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000219097.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000219097.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000219097.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000219097.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000219097.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000219097.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000219097.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000563306.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000563306.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000563306.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000563306.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000563306.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000563306.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000563306.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000563306.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000563306.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000563306.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000563599.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000563599.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000563599.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000563599.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000563599.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000563599.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000563599.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000566860.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000566860.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000566860.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000566860.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000566860.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000566860.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000566860.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000566860.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000566860.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000566860.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000566860.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000566860.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000567000.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000567000.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000567000.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000567000.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000567000.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000567000.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000567000.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000567000.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000568364.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000568364.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000568364.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000568364.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000568364.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000568364.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000568364.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000568364.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000568364.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000569239.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000569239.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000569239.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000569239.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000569239.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000569239.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000570260.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000570260.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000570260.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000570260.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000570260.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000570260.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000575571.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000575571.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000575571.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000575571.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000575571.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000575571.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000575571.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 ENST00000575571.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NM_014321.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NM_014321.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NM_014321.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NM_014321.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NM_014321.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NM_014321.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NM_014321.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NM_014321.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NM_014321.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NM_014321.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NM_014321.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NM_014321.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NR_037620.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NR_037620.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NR_037620.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NR_037620.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NR_037620.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NR_037620.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NR_037620.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NR_037620.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NR_037620.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NR_037620.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NR_037620.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NR_037620.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007718.hg.1 NR_037620.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009870.hg.1 ENST00000529323.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009870.hg.1 ENST00000529323.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006791.hg.1 ENST00000623071.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006791.hg.1 ENST00000623071.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006791.hg.1 ENST00000623071.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006791.hg.1 ENST00000624346.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200009108.hg.1 ENST00000625200.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010748.hg.1 ENST00000624108.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline