| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07016179.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07016179.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07016179.hg.1 |
TC07001050.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000460332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000460332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000460332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000460332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000469271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000469271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000469271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000469271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000469271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000469271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000469271.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000472491.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000472491.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000472491.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000472491.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000472491.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000475176.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000475176.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000475176.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000475176.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000475176.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000475176.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000485661.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000485661.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000485661.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000485661.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000486131.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000486131.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000486131.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000486131.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000489850.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000489850.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000489850.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
ENST00000489850.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07016175.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07016175.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07016175.hg.1 |
TC07001050.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700170156.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170156.hg.1 |
ENST00000475176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170156.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700170156.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170156.hg.1 |
ENST00000475176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170156.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700170156.hg.1 |
TC0700009860.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700170160.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170160.hg.1 |
ENST00000469271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170160.hg.1 |
ENST00000472491.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170160.hg.1 |
ENST00000475176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170160.hg.1 |
ENST00000489850.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170160.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700170160.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170160.hg.1 |
ENST00000469271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170160.hg.1 |
ENST00000472491.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170160.hg.1 |
ENST00000475176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170160.hg.1 |
ENST00000489850.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170160.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700170160.hg.1 |
TC0700009860.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000275820.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000460332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000460332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000460332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000460332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000469271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000469271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000469271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000469271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000469271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000469271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000469271.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000472491.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000472491.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000472491.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000472491.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000472491.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000475176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000475176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000475176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000475176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000475176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000475176.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000485661.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000485661.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000485661.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000485661.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000486131.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000486131.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000486131.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000486131.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000489850.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000489850.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000489850.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
ENST00000489850.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009860.hg.1 |
NM_138400.1 |
Refseq |